Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • Occludin Antibodies
          Now also available in recombinant format (Product # 740006M) and additional conjugated formats

          Invitrogen

          Occludin Monoclonal Antibody (OC-3F10)

          Advanced Verification
          92 Published Figures
          591 References
          View all (55) Occludin antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite Occludin Monoclonal Antibody (OC-3F10)

          Additional Information:

          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (4)
          • Published Figures (92)
          • Advanced Verification (2)
          Occludin Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Occludin Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 98

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Occludin Antibody (33-1500) in ICC/IF

          Immunofluorescent analysis of Occludin was done on 70% confluent log phase MDCK cells. The cells were fixed with 4% paraformaldehyde for 15 minutes; permeabilized with 0.25% Triton™ X-100 for 10 minutes followed by blocking with 5% BSA for 1 hour at room temperature. The cells were incubated with Occludin Mouse Monoclonal Antibody (Product # 33-1500) at 1 µg/mL in 1% BSA and incubated for 3 hours at room temperature and then labeled with Alexa Fluor® 488 Rabbit Anti-Mouse IgG Secondary Antibody (Product # A-11059) at a dilution of 1:400 fo... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Occludin Antibody in Immunocytochemistry (ICC/IF)
          Occludin Antibody in Immunocytochemistry (ICC/IF)
          Occludin Antibody in Western Blot (WB)
          Occludin Antibody in Western Blot (WB)
          Occludin Antibody in Immunocytochemistry (ICC/IF)
          Occludin Antibody in Immunocytochemistry (ICC/IF)
          Occludin Antibody in Immunocytochemistry (ICC/IF)
          Occludin Antibody in Immunocytochemistry (ICC/IF)
          Occludin Antibody in Immunocytochemistry (ICC/IF)
          Occludin Antibody in Immunocytochemistry (ICC/IF)
          Occludin Antibody in Immunocytochemistry (ICC/IF)
          Occludin Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          Occludin Antibody in Immunocytochemistry (ICC/IF)
          Occludin Antibody in Immunocytochemistry (ICC/IF)
          Occludin Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          Occludin Antibody in Immunocytochemistry (ICC/IF)
          Occludin Antibody in Immunocytochemistry, Immunoprecipitation (ICC/IF, IP)
          Occludin Antibody in Immunocytochemistry (ICC/IF)
          Occludin Antibody in Immunocytochemistry (ICC/IF)
          Occludin Antibody in Immunocytochemistry (ICC/IF)
          Occludin Antibody in Immunohistochemistry, Immunocytochemistry (IHC, ICC/IF)
          Occludin Antibody in Immunocytochemistry (ICC/IF)
          Occludin Antibody in Immunocytochemistry (ICC/IF)
          Occludin Antibody in Immunocytochemistry (ICC/IF)
          Occludin Antibody in Immunocytochemistry (ICC/IF)
          Occludin Antibody in Immunocytochemistry (ICC/IF)
          Occludin Antibody in Immunocytochemistry (ICC/IF)
          Occludin Antibody in Immunohistochemistry, Immunocytochemistry (IHC, ICC/IF)
          Occludin Antibody in Immunocytochemistry (ICC/IF)
          Occludin Antibody in Immunocytochemistry (ICC/IF)
          Occludin Antibody in Immunocytochemistry (ICC/IF)
          Occludin Antibody in Immunocytochemistry (ICC/IF)
          Occludin Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          Occludin Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          Occludin Antibody in Immunocytochemistry (ICC/IF)
          Occludin Antibody in Immunocytochemistry (ICC/IF)
          Occludin Antibody in Western Blot, Immunocytochemistry, Flow Cytometry (WB, ICC/IF, Flow)
          Occludin Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          Occludin Antibody in Immunocytochemistry (ICC/IF)
          Occludin Antibody in Immunocytochemistry (ICC/IF)
          Occludin Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          Occludin Antibody in Immunocytochemistry (ICC/IF)
          Occludin Antibody in Immunocytochemistry (ICC/IF)
          Occludin Antibody in Immunocytochemistry (ICC/IF)
          Occludin Antibody in Immunocytochemistry (ICC/IF)
          Occludin Antibody in Immunocytochemistry (ICC/IF)
          Occludin Antibody in Immunocytochemistry (ICC/IF)
          Occludin Antibody in Immunocytochemistry (ICC/IF)
          Occludin Antibody in Immunocytochemistry (ICC/IF)
          Occludin Antibody in Immunocytochemistry (ICC/IF)
          Occludin Antibody in Immunohistochemistry, Immunocytochemistry (IHC, ICC/IF)
          Occludin Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          Occludin Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          Occludin Antibody in Immunocytochemistry (ICC/IF)
          Occludin Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          Occludin Antibody in Immunocytochemistry (ICC/IF)
          Occludin Antibody in Immunocytochemistry (ICC/IF)
          Occludin Antibody in Immunocytochemistry (ICC/IF)
          Occludin Antibody in Immunocytochemistry (ICC/IF)
          Occludin Antibody in Immunocytochemistry (ICC/IF)
          Occludin Antibody in Immunocytochemistry (ICC/IF)
          Occludin Antibody in Immunocytochemistry (ICC/IF)
          Occludin Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          Occludin Antibody in Immunocytochemistry (ICC/IF)
          Occludin Antibody in Immunocytochemistry (ICC/IF)
          Occludin Antibody in Immunocytochemistry (ICC/IF)
          Occludin Antibody in Immunocytochemistry (ICC/IF)
          Occludin Antibody in Immunocytochemistry (ICC/IF)
          Occludin Antibody in Immunocytochemistry (ICC/IF)
          Occludin Antibody in Immunocytochemistry (ICC/IF)
          Occludin Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          Occludin Antibody in Immunocytochemistry (ICC/IF)
          Occludin Antibody in Immunocytochemistry (ICC/IF)
          Occludin Antibody in Immunocytochemistry (ICC/IF)
          Occludin Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Occludin Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Occludin Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Occludin Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Occludin Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Occludin Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Occludin Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Occludin Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Occludin Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Occludin Antibody in Western Blot, Immunohistochemistry (Frozen) (WB, IHC (F))
          Occludin Antibody in Western Blot, Immunohistochemistry (Frozen) (WB, IHC (F))
          Occludin Antibody in Western Blot, Immunohistochemistry (Frozen) (WB, IHC (F))
          Occludin Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Occludin Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Occludin Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Occludin Antibody in Immunohistochemistry (IHC)
          Occludin Antibody in Immunohistochemistry (IHC)
          Occludin Antibody in Immunohistochemistry (IHC)
          Occludin Antibody in Immunohistochemistry, Immunoprecipitation (IHC, IP)
          Occludin Antibody in Immunohistochemistry (IHC)
          Occludin Antibody in Immunohistochemistry (IHC)
          Occludin Antibody in Western Blot, Immunohistochemistry (WB, IHC)
          Occludin Antibody
          Occludin Antibody

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Occludin Monoclonal Antibody (OC-3F10)

          Product Details

          33-1500

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          0.1-1.0 µg/mL
          View 225 publications 225 publications

          Immunohistochemistry (IHC)

          -
          View 91 publications 91 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          -
          View 12 publications 12 publications

          Immunohistochemistry (Frozen) (IHC (F))

          -
          View 28 publications 28 publications

          Immunocytochemistry (ICC/IF)

          2-3 µg/mL
          View 182 publications 182 publications

          Flow Cytometry (Flow)

          -
          View 4 publications 4 publications

          ELISA (ELISA)

          0.1-1.0 µg/mL
          View 2 publications 2 publications

          Immunoprecipitation (IP)

          -
          View 5 publications 5 publications

          Proximity Ligation Assay (PLA) (PLA)

          -
          View 1 publication 1 publication

          Miscellaneous PubMed (Misc)

          -
          View 41 publications 41 publications
          Product Specifications

          Species Reactivity

          Dog, Human, Mouse, Rat

          Published species

          Bacteria, Bovine, C. elegans, Chicken, Cynomolgus monkey, Dog, Guinea pig, Human, Mouse, Pig, Rabbit, Rat, Rhesus monkey, Sheep, Tag

          Host/Isotype

          Mouse / IgG1, kappa

          Class

          Monoclonal

          Type

          Antibody

          Clone

          OC-3F10

          Immunogen

          GST fusion protein consisting of the C-terminal region (~150aa) of human occludin.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated
          • Alexa Fluor 488 
          • Alexa Fluor 594 
          • FITC
          • HRP
          • Request custom conjugation

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Amount

          100 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          0.1% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2533101

          Target Information

          Occludin is a 65 kDa protein that can exist in a variety of phosphorylated forms, ranging up to approximately 82 kDa. Occludin is thought to be involved in regulating both the localization and the function of occludin. Polyunsaturated fatty acids are known to up-regulate occludin expression, increasing the transendothelial cell resistance and reducing the cellular permeability to large molecules. The level of occludin varies greatly depending on tissue; in brain tissue, occludin is highly and continuously expressed at cell-cell contact sites, whereas non-neural tissues show lower expression and discontinuous distribution. Overall structural features of the occludin protein are highly conserved in all the species examined. Under-expression of tight junction proteins, including occludin, is a key molecular abnormality responsible for the increased permeability of tumor endothelial tight junctions, which contributes to brain tumor edemas.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: phosphatase 1, regulatory subunit 115; tight junction protein; tight junction structural protein; unnamed protein product

          View more View less

          Gene Aliases: BLCPMG; Ocl; PPP1R115; PTORCH1

          View more View less

          UniProt ID: (Human) Q16625, (Dog) Q28269, (Mouse) Q61146, (Rat) Q6P6T5

          View more View less

          Entrez Gene ID: (Human) 100506658, (Dog) 403844, (Mouse) 18260, (Rat) 83497

          View more View less

          Function(s)
          protein binding protein domain specific binding general transcription factor
          Process(es)
          positive regulation of lamellipodium assembly positive regulation of gene expression negative regulation of gene expression epithelial cell migration regulation of glucose transport positive regulation of microtubule polymerization maintenance of blood-brain barrier cell-cell junction organization positive regulation of glucose import protein-containing complex assembly bicellular tight junction assembly positive regulation of wound healing tight junction organization protein localization to cell leading edge positive regulation of maintenance of permeability of blood-brain barrier response to interleukin-18 sensory perception of sound response to ethanol establishment of endothelial barrier cellular response to tumor necrosis factor response to Gram-positive bacterium
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-5bc99cc877-qh5rm:80/100.66.76.16:80.
          git-commit: a70b05477dc28cec55d7eafa2ce360191b99e591
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.58.0-2026.09.99-1.0