BCIP Substrate Powder (5-bromo-4-chloro-3'-indolylphosphate p-toluidine) - FAQs
View additional product information for BCIP Substrate Powder (5-bromo-4-chloro-3'-indolylphosphate p-toluidine) - FAQs (34040)
1 product FAQs found