1-Step™ PNPP Substrate Solution, 100 mL - FAQs
1-Step™ PNPP Substrate Solution, 100 mL - FAQs
View additional product information for PNPP Substrates - FAQs (34047, 34045, 37620, 37621)
1 product FAQs found
What alkaline phosphatase substrates do you offer?