Neuron Isolation Enzyme (with papain) for Pierce™ Primary Cell Isolation Kits
Thermo Scientific™

Neuron Isolation Enzyme (with papain) for Pierce™ Primary Cell Isolation Kits

Neuronal Isolation Enzyme (with papain) is a tissue-specific dissociation reagent for the isolation of primary cortical and hippocampal neurons fromRead more
Have Questions?
Catalog NumberQuantity
882851 Vial
Catalog number 88285
Price (USD)
79.65
Online exclusive
86.75
Save 7.10
Each
Quantity:
1 Vial
Request bulk or custom format
Price (USD)
79.65
Online exclusive
86.75
Save 7.10
Each
Neuronal Isolation Enzyme (with papain) is a tissue-specific dissociation reagent for the isolation of primary cortical and hippocampal neurons from mouse/rat embryonic brain tissue and is a component of the Thermo Scientific™ Pierce™ Primary Neuron Isolation Kit (Product No. 88280).

Papain has been demonstrated to be more effective and less harsh than other proteases for tissue dissociation. When used with Cardiomyocyte Isolation Enzyme 2 (with thermolysin), it improves digestion efficiency and cell yield of primary cardiomyocytes.

Papain Information:

• Source:Papaya latex
• Molecular mass: 23kDa
• Optimal pH: 6.0-7.0

More Product Data
• An efficient method for the isolation of highly functional primary neurons

Related Products
Pierce™ Primary Neuron Isolation Kit
Neuronal Culture Medium for Pierce™ Primary Cell Isolation Kits
Neuronal Culture Media Supplement (50X) for Pierce™ Primary Cell Isolation Kits
N-PER™ Neuronal Protein Extraction Reagent
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell TypeNeuronal Cells (Pre-Natal), Primary Cells
Quantity1 Vial
Target SpeciesRat, Mouse
FormSolid
Product LinePierce
Product TypeCell Isolation Enzyme
Unit SizeEach
Contents & Storage
Stability: 1 year at -20°C
Storage of Reconstituted Enzyme: -20°C for 6 months and up to two freeze-thaw cycles, or 1 week at 4°C. Enzyme use after prolonged storage may result in poor performance.