Product Image
Applied Biosystems™

GeneTitan™ Hybridization, Wash, and Stain Kit for WT Array Plates

The GeneTitan™ Hybridization, Wash, and Stain Kit for WT Arrays contains pre-formulated solutions to process WT array plates on theRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
905102384 Reactions
90162296 Reactions
Catalog number 905102
Price (USD)
-
Quantity:
384 Reactions
The GeneTitan™ Hybridization, Wash, and Stain Kit for WT Arrays contains pre-formulated solutions to process WT array plates on the GeneTitan™ System. The reagents are formulated into the fewest number of individual components possible, reducing preparation and handling steps. With enhanced performance, fewer required reagents, and a simple protocol, target preparation has never been so easy.

The GeneTitan Hybridization, Wash, and Stain Kit for WT Arrays includes two boxes:

The first box contains sufficient amounts of the hybridization and stain components to process one 96-array plate, four 24-array plates, or six 16-array plates. The hybridization components include the solutions required for preparation of the hybridization cocktail containing the target. The stain components include Stain Cocktail 1 and 3, Stain Cocktail 2, and the Array Holding Buffer. These solutions are used during the processing of the arrays in the GeneTitan System.

The second box includes Wash Buffer A and Wash Buffer B, also required for the processing of the arrays. There is sufficient Wash Buffer A and B to process one plate in any format. For each additional plate that is processed, an additional Wash Buffer A and B module must be purchased.

The Stain Cocktail 1 and 3, Stain Cocktail 2, Array Holding Buffer, and Wash Buffers A and B are ready-to-use solutions. The hybridization cocktail is prepared by combining the Hybridization Mix and nuclease-free water with the target prepared for the specific target labeling assay (IVT or WT).

The Applied Biosystems GeneTitan Hybridization, Wash, and Stain Kit for WT Arrays should always be stored at 2 to 8° C.

Related Links
GeneTitan™ Hybridization, Wash, and Stain Kit for 3' IVT Arrays
GeneTitan™ Wash Buffers A and B Module
GeneChip™ Hybridization Control Kit
B2 Control Oligo (3nM)
GeneChip™ miRNA 4.1 24-Array Plate and Trays
GeneChip™ miRNA 4.1 96-Array Plate and Trays
GeneTitan™ miRNA Array Holding Buffer
GeneChip™ Hybridization Control Kit
GeneChip™ miRNA 3.1 24-Array Plate and Trays
GeneChip™ miRNA 3.1 96-Array Plate and Trays
FlashTag™ Biotin HSR RNA Labeling Kits
FlashTag™ Biotin HSR RNA Labeling Kits
GeneTitan™ Hybridization Module for miRNA Plates
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Transcriptome Profiling
Quantity384 Reactions
Reagent TypeHybridization, Wash, Stain
TypeHybridization, Wash, Stain Kit
Format24-array Plate, 96-array Plate
Unit SizeEach