Product Image
Invitrogen™
iBright™ Imaging System Sample Stage/Turntable
Used for sample placement and auto-alignment in the iBright imaging systems
Have Questions?
Catalog NumberQuantity
A338291 Ea.
Catalog number A33829
Price (USD)
628.00
Each
Quantity:
1 Ea.
Price (USD)
628.00
Each
The iBright Imaging System Sample Stage/Turntable is used for sample placement and auto-alignment in the iBright imaging systems (CL750, CL1000, CL1500, FL1000, FL1500). It is made available as a replacement or additional sample stage for these systems. If the iBright imaging system is being used extensively for imaging and band excision from DNA gels, it is recommended that users have a separate Sample Stage to dedicate to this application.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Chemiluminescent Western Blot Imaging, DNA Gel Imaging, Protein Gel Imaging
Product LineiBright
Quantity1 Ea.
Shipping ConditionRoom Temperature
Product TypeImaging System
Unit SizeEach
Frequently asked questions (FAQs)
Why do the iBright Imaging Systems use green LED illumination for DNA imaging?
The iBright Imaging Systems utilize a transilluminator based on green LEDs, which effectively excite popular DNA dyes such ethidium bromide and SYBR Green dye and offer several additional benefits:
- No harmful UV rays: While UV light effectively excites many fluorescent dyes and stains, UV light is a health hazard. Further, prolonged exposure to UV light can damage DNA samples, and compromise the integrity of samples to be used for downstream applications, such as subcloning.
- No mercury waste: UV transilluminator bulbs may contain mercury, a hazardous substance, and therefore require special care for handling and disposal.
- Longer lifetime: LED bulbs have a substantially longer real-time life than fluorescent UV bulbs, which can add up to considerable cost savings over the lifetime of the instrument.
For additional information on iBright functionality, visit this page.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.
How large is the sample imaging area on the iBright Imaging Systems?
The imaging area of the iBright Imaging system is 22.5 x 18.0 cm. This gives ample space for imaging 4 mini blots (or 2 midi blots) simultaneously.
For more information on iBright features and capabilities, visit this page.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.
What is the weight of the Thermal Printer for iBright Imaging Systems?
The Thermal Printer for iBright Imaging Systems weighs approximately 5.5 lbs (2.5 kg).
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.
What are the dimensions of the Thermal Printer for iBright Imaging Systems?
Here are the dimensions of the Thermal Printer for iBright Imaging Systems:
Approximately 3.46 x 6.06 x 9.45 inches (88 x 154 x 240 mm).
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.
What is the power consumption of the Thermal Printer for iBright Imaging Systems?
In stand-by mode, the power consumption of the Thermal Printer for iBright Imaging Systems is 10 Watts, and during the printing process, it is 115 Watts.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.