Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • CD85j (ILT2) Antibodies

          Invitrogen

          CD85j (ILT2) Monoclonal Antibody (HP-F1), NovaFluor™ Blue 610-70S, eBioscience™

          View all (51) CD85j (ILT2) antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite CD85j (ILT2) Monoclonal Antibody (HP-F1), NovaFluor™ Blue 610-70S, eBioscience™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (2)
          CD85j (ILT2) Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          CD85j (ILT2) Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.

          FIGURE: 1 / 2

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          CD85j (ILT2) Antibody (H043T02B06-A) in Flow

          Normal human peripheral blood cells were unstained (left) or stained with CD85j (ILT2) Monoclonal Antibody, NovaFluor Blue 610-70S (right). All cells were co-stained with CD11b Monoclonal Antibody, Super Bright 780 (Product # 78-0118-42). Total viable cells in the monocyte gate were used for analysis, as determined by LIVE/DEAD Blue (Product # L34962). Data was acquired on a 5-laser Cytek Aurora and unmixed with autofluorescence extraction. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          CD85j (ILT2) Antibody in Flow Cytometry (Flow)
          CD85j (ILT2) Antibody in Flow Cytometry (Flow)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          H043T02B06-A

          Applications
          Tested Dilution
          Publications

          Flow Cytometry (Flow)

          5 µL (0.6 µg)/test
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG1, kappa

          Class

          Monoclonal

          Type

          Antibody

          Clone

          HP-F1

          Conjugate

          NovaFluor™ Blue 610-70S NovaFluor™ Blue 610-70S NovaFluor™ Blue 610-70S
          • Alexa Fluor 488 
          • Alexa Fluor 700 
          • APC 
          • APC-eFluor 780
          • Brilliant UV 737
          • eFluor 450
          • FITC
          • Functional Grade
          • NovaFluor Blue 660-120S
          • NovaFluor Blue 690
          • NovaFluor Red 700
          • NovaFluor Red 725
          • NovaFluor Yellow 570
          • NovaFluor Yellow 590
          • NovaFluor Yellow 610
          • NovaFluor Yellow 660
          • PE 
          • PE-Cyanine7
          • PE-eFluor 610
          • PerCP-eFluor 710
          • Super Bright 436
          • Super Bright 600
          • Request custom conjugation

          Excitation/Emission Max

          492/616 nm View spectra spectra

          Form

          Liquid

          Concentration

          0.6 µg/Test

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.2

          Contains

          0.09% sodium azide

          Storage conditions

          4°C, store in dark, DO NOT FREEZE!

          RRID

          AB_3098520

          Product Specific Information

          Description
          The monoclonal antibody HP-F1 recognizes CD85j, also known as ILT2, LILRB1, and LIR1. The monoclonal antibody HP-F1 has been shown to reduce the amount of CD16- dependent cytolytic activity of functional NK cells.

          This product contains 1 vial of NovaFluor conjugate and 1 vial of CellBlox Plus Blocking Buffer.

          Applications Tested
          This HP-F1 antibody has been pre-diluted and tested by flow cytometric analysis of normal human peripheral blood cells. This may be used at 5 µL (0.25 µg) per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.

          Master mixes
          • Master mixes of NFs should be made at 2-8 °C and may be made up to 4 hours ahead of time.
          • We do not recommend storing master mixes containing NovaFluor conjugates overnight or longer.

          Whole Blood compatibility
          • When utilizing whole blood (as opposed to density-gradient-purified PBMC), we recommend lysing red blood cells in bulk prior to staining with NovaFluor conjugates.
          • See the Bulk Lysis of Human Whole Blood protocol here.
          • Staining of whole blood with NovaFluor conjugates followed by lysis of red blood cells may result in higher-than-expected background staining.

          Viability dye compatibility
          • NovaFluor dyes are not compatible with DNA intercalating viability dyes.
          • Do not use viability dyes such as propidium iodide, 7-actinomycin D (7-AAD) and DAPI. Invitrogen LIVE/DEAD Fixable Dead Cell stains are recommended for use with NovaFluor dyes.

          CellBlox Plus Blocking Buffer
          • This NovaFluor conjugate comes with CellBlox Plus Blocking Buffer (for 25 µg or 25 test products, Cat. No. C001T02F01; for 100 µg or 100 test, Cat. No. C001T03F01), essential for optimal staining.
          • Use CellBlox Plus Blocking Buffer in all experiments with NovaFluor conjugates.
          • Add 5 µL per sample to antibody cocktails/master mixes (regardless of how many Novafluor-conjugated antibodies are present) before combining with cells.
          • CellBlox Plus Blocking Buffer is compatible with either Super Bright Complete Blocking Buffer or Brilliant Stain Buffer and can be used in antibody cocktails/master mixes with those reagents.
          • For single-color controls, use 5 µL of CellBlox Plus Blocking Buffer per 100 µL of cell sample (10^3 to 10^8 cells).

          NovaFluor conjugates are based on Phiton technology utilizing novel fluorophore-containing nucleic acid dye structures that allow for engineered fluorescent signatures with consideration for spillover and spread impacts. Learn more

          Excitation: 492 nm; Emission: 616 nm; Laser: 488 nm (Blue) Laser

          Target Information

          CD85j, also known as a member of the leukocyte immunoglobulin-like receptor (LIR) family, is a single transmembrane glycoprotein with a long cytoplasmic domain containing four immunoreceptor tyrosine-based inhibitory motifs (ITIMs). These motifs signal through interactions with SHP-1, transducing negative signals that inhibit immune response stimulation. CD85j is expressed on myeloid cells such as monocytes and dendritic cells, as well as on subsets of lymphoid cells including NK, T, and B cells. The expression of CD85j is correlated with leukemias such as acute lymphoblastic leukemia (ALL) and chronic lymphocytic leukemia (CLL). On CD8+ cells, CD85j expression is associated with effector cell function and plays a significant role in viral infections, including HIV, Epstein-Barr virus, and cytomegalovirus (CMV). CD85j binds to MHC class I molecules, including HLA-G, A, F, B27, E, and F, on antigen-presenting cells. The receptor helps control inflammatory responses and cytotoxicity, focusing the immune response and limiting autoreactivity. CD85j belongs to the subfamily B class of LIR receptors, which contain two or four extracellular immunoglobulin domains, a transmembrane domain, and multiple transcript variants encoding different isoforms have been identified for this gene. Its gene is located in a cluster at chromosomal region 19q13.4.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          How to use the Panel Builder

          Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: CD85 antigen-like family member J; Ig-like transcript 2; ILT2a; allelic form of ILT2; ILT2b; alternatively spliced form of ILT2; ILT2c; alternatively spliced form of ILT2; immunoglobulin heavy chain variable region; leucocyte Ig-like receptor B1; leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), member 1; LIR-1; monocyte/macrophage immunoglobulin-like receptor 7; myeloid inhibitory receptor 7; unnamed protein product

          View more View less

          Gene Aliases: CD85J; ILT-2; ILT2; LIR-1; LIR1; MIR-7; MIR7; PIR-B; PIRB

          View more View less

          UniProt ID: (Human) Q8NHL6

          View more View less

          Entrez Gene ID: (Human) 10859

          View more View less

          Function(s)
          beta-amyloid binding protein binding protein phosphatase 1 binding MHC class Ib protein complex binding MHC class Ib protein binding HLA-A specific inhibitory MHC class I receptor activity HLA-B specific inhibitory MHC class I receptor activity MHC class I receptor activity MHC class Ib receptor activity inhibitory MHC class I receptor activity SH2 domain binding MHC class I protein binding protein homodimerization activity
          Process(es)
          negative regulation of T cell mediated cytotoxicity positive regulation of defense response to virus by host T cell proliferation involved in immune response negative regulation of cytokine production involved in immune response immune response-inhibiting cell surface receptor signaling pathway Fc receptor mediated inhibitory signaling pathway signal transduction cell surface receptor signaling pathway response to virus positive regulation of gene expression negative regulation of serotonin secretion receptor internalization negative regulation of interferon-beta production negative regulation of interferon-gamma production negative regulation of interleukin-10 production negative regulation of interleukin-12 production negative regulation of tumor necrosis factor production positive regulation of interferon-gamma production negative regulation of mononuclear cell proliferation negative regulation of T cell proliferation positive regulation of apoptotic process negative regulation of cell cycle negative regulation of endocytosis positive regulation of transcription from RNA polymerase II promoter negative regulation of natural killer cell mediated cytotoxicity negative regulation of alpha-beta T cell activation defense response to virus negative regulation of calcium ion transport positive regulation of macrophage cytokine production cellular response to lipopolysaccharide negative regulation of transforming growth factor beta production dendritic cell differentiation interleukin-10-mediated signaling pathway negative regulation of dendritic cell apoptotic process negative regulation of CD8-positive, alpha-beta T cell activation negative regulation of T cell activation via T cell receptor contact with antigen bound to MHC molecule on antigen presenting cell negative regulation of dendritic cell differentiation negative regulation of osteoclast development
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-76c4fffd86-jvpnf:80/100.66.74.151:80.
          git-commit: bdc067a1e9e92d056899a2dda64af631988aa8a1
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.54.0-Offline