Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • RLTPR Antibodies

          Invitrogen

          RLTPR Monoclonal Antibody (EM-53)

          View all (9) RLTPR antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite RLTPR Monoclonal Antibody (EM-53)

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          RLTPR Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          RLTPR Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          RLTPR Antibody (MA5-28632) in Flow

          Separation of RLTPR transfected cells stained using anti-human RLTPR (EM-53) purified Monoclonal antibody (Product # MA5-28632) (GAM FITC, concentration in sample 9 µg/mL, red-filled) from RLTPR transfected cells stained using mouse IgG1 isotype control (MOPC-21) purified antibody (GAM FITC, concentration in sample 9 µg/mL, black-dashed) in flow cytometry analysis (intracellular staining). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          RLTPR Antibody in Flow Cytometry (Flow)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          RLTPR Monoclonal Antibody (EM-53)

          Product Details

          MA5-28632

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          Assay-dependent
          -

          Flow Cytometry (Flow)

          1-4 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human, Mouse

          Host/Isotype

          Mouse / IgG1, kappa

          Class

          Monoclonal

          Type

          Antibody

          Clone

          EM-53

          Immunogen

          Murine Rltpr
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated
          • APC 
          • PE 
          • Request custom conjugation

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          15mM sodium azide

          Storage conditions

          4°C, do not freeze

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2745591

          Product Specific Information

          This antibody recognizes RLTPR / CARMIL2, an intracellular protein playing a role in actin filament elongation.

          Target Information

          RLTPR/CARMIL2 (RGD motif, leucine rich repeats, tropomodulin domain and proline-rich containing; capping protein regulator and myosin 1 linker 2), also known as LRRC16C, is a cytosolic protein, which with high affinity binds CAPZA2 (capping protein muscle actin Z-line alpha 2) and decreases CAPZA2 affinity for actin barbed ends. RLTPR/CARMIL2 increases the rate of actin filament elongation from seeds in the presence of CAPZA2, however, seems unable to nucleate filaments. Its interaction with CAPZA2 is essential for lamellipodial protrusion and cell translocation. RLTPR/CARMIL2 is crucial for T cell costimulation via CD28 and this property seems to be independent on its actin-uncapping function. The lack of functional RLTPR/CARMIL2 molecules impeded the differentiation toward Th1 and Th17 fates of both human and murine CD4+ T cells and leads to combined immunodeficiency. Expression of RLTPR/CARMIL2 was also detected in human and murine B cells, but it seems not to be involved in BCR-mediated signaling.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: cytoskeleton protein; F-actin-uncapping protein RLTPR; leucine rich repeat containing 16C; leucine-rich repeat-containing protein 16C; RGD motif, leucine rich repeats, tropomodulin domain and proline-rich containing; RGD, leucine-rich repeat, tropomodulin and proline-rich containing protein; RGD, leucine-rich repeat, tropomodulin and proline-rich-containing protein; unnamed protein product

          View more View less

          Gene Aliases: CARMIL2b; CG1399-PB; D130029J02Rik; Gm585; IMD58; LRRC16C; RLTPR

          View more View less

          UniProt ID: (Human) Q6F5E8

          View more View less

          Entrez Gene ID: (Human) 146206, (Mouse) 234695

          View more View less

          Function(s)
          phospholipid binding macromolecular complex binding protein binding
          Process(es)
          establishment or maintenance of cell polarity positive regulation of lamellipodium assembly cell migration positive regulation of cell migration regulation of Arp2/3 complex-mediated actin nucleation wound healing, spreading of cells actin filament network formation establishment or maintenance of monopolar cell polarity positive regulation of extracellular matrix disassembly positive regulation of ruffle assembly positive regulation of lamellipodium organization negative regulation of barbed-end actin filament capping positive regulation of interferon-gamma production positive regulation of interleukin-2 production positive regulation of T cell proliferation establishment of protein localization positive regulation of regulatory T cell differentiation thymus development homeostasis of number of cells T cell receptor signaling pathway
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5799cd5976-2rlvt:80/100.66.78.131:80.
          git-commit: e30ab741173a2eb62411af663260592d4bdb9634
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.55.0-2026.07.30-1.0