Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • PTEN Antibodies

          Invitrogen

          Phospho-PTEN (Ser380) Recombinant Rabbit Monoclonal Antibody (PTENS380-NA9)

          View all (23) PTEN antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Phospho-PTEN (Ser380) Recombinant Rabbit Monoclonal Antibody (PTENS380-NA9)

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation'] || 'Additional Information'}}: {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (3)
          Phospho-PTEN (Ser380) Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          Phospho-PTEN (Ser380) Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.

          FIGURE: 1 / 3

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Phospho-PTEN (Ser380) Antibody (MA5-37050) in Flow

          Flow cytometry of Phospho-PTEN (Ser380) using A431 cells. Samples were incubated in monoclonal Phospho-PTEN (Ser380) antibody (Product # MA5-37050) with a dilution of 0.05 µg/mL. Images correspond with the following conditions: negative control (light blue), untreated (red), treated with EGF (green), untreated and blocked with phospho-peptide (black), EGF and blocked with phospho peptide (gold), untreated a... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Phospho-PTEN (Ser380) Antibody in Flow Cytometry (Flow)
          Phospho-PTEN (Ser380) Antibody in Flow Cytometry (Flow)
          Phospho-PTEN (Ser380) Antibody in Flow Cytometry (Flow)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Phospho-PTEN (Ser380) Recombinant Rabbit Monoclonal Antibody (PTENS380-NA9)

          Product Details

          MA5-37050

          Applications
          Tested Dilution
          Publications

          Flow Cytometry (Flow)

          1-1,000 ng/mL
          -
          Product Specifications

          Species Reactivity

          Human, Mouse

          Host/Isotype

          Rabbit / IgG, kappa

          Expression System

          HEK293 cells

          Class

          Recombinant Monoclonal

          Type

          Antibody

          Clone

          PTENS380-NA9

          Immunogen

          A synthetic phospho-peptide corresponding to residues surrounding Ser380 of human phospho PTEN.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated
          • APC 
          • FITC
          • PE 
          • Request custom conjugation

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Amount

          100 µg

          Purification

          Protein A/G

          Storage buffer

          PBS with 0.10% BSA, 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2896985

          Target Information

          PTEN was identified as a tumor suppressor that is mutated in a large number of cancers at high frequency. The PTEN gene is a phosphatidylinositol-3, 4, 5-trisphosphate 3-phosphatase. It contains a tensin like domain as well as a catalytic domain similar to that of the dual specificity protein tyrosine phosphatases. Unlike most of the protein tyrosine phosphatases, this protein preferentially dephosphorylates phosphoinositide substrates. PTEN negatively regulates intracellular levels of phosphatidylinositol-3, 4, 5-trisphosphate in cells and functions as a tumor suppressor by negatively regulating AKT/PKB signaling pathway. PTEN plays a role as a key modulator of the AKT-mTOR signaling pathway controlling the tempo of the process of newborn neurons integration during adult neurogenesis, including correct neuron positioning, dendritic development and synapse formation. Also, PTEN may be a negative regulator of insulin signaling and glucose metabolism in adipose tissue. The nuclear monoubiquitinated form possesses greater apoptotic potential, whereas the cytoplasmic nonubiquitinated form of PTEN induces less tumor suppressive ability.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: inositol polyphosphate 3-phosphatase; mitochondrial phosphatase and tensin protein alpha; mitochondrial PTENalpha; MMAC1 phosphatase and tensin homolog deleted on chromosome 10; mutated in multiple advanced cancers 1; N-terminal extended isoform of PTEN translated from an alternative ATT initiation codon in the 5'UTR; N-terminal extended PTEN isoform; uses an alternative ATT initiation codon in PTEN 5'UTR; phosphatidylinositol-3,4,5-trisphosphate3-phosphatase; phosphatase and tensin; phosphatase and tensin-like protein; phosphatidylinositol 3; phosphatidylinositol 3,4,5-trisphosphate 3-phosphatase and dual-specificity protein phosphatase PTEN isoform 5; phosphatidylinositol 3,4,5-trisphosphate 3-phosphatase and dual-specificity protein phosphatase PTEN isoform 6; phosphatidylinositol 3,4,5-trisphosphate 3-phosphatase and dual-specificity protein phosphatase PTEN isoform 7; phosphatidylinositol 3,4,5-trisphosphate 3-phosphatase and dual-specificity protein phosphatase PTEN, isoform 21; phosphatidylinositol 3,4,5-trisphosphate 3-phosphatase and dual-specificity protein phosphatase PTEN, isoform 22; phosphatidylinositol-3; phosphatidylinositol-3,4,5-trisphosphate 3-phosphatase and dual-specificity protein phosphatase PTEN; protein tyrosine phosphatase; protein tyrosine phosphatase and tensin homolog; PTEN; pten tumor suppressor; PTEN variant PTEN-K267Rfs*9; PTEN variant PTEN-L247*; PTEN variant PTEN-R234Afs*11; PTENepsilon; TEP1; transcript variant 1; unnamed protein product

          View more View less

          Gene Aliases: 10q23del; 2310035O07Rik; A130070J02Rik; B430203M17Rik; BZS; CWS1; DEC; GLM2; MHAM; MMAC1; PTEN1; PTENbeta; PTENgama; TEP1

          View more View less

          UniProt ID: (Human) P60484, (Mouse) O08586

          View more View less

          Entrez Gene ID: (Human) 5728, (Mouse) 19211

          View more View less

          Function(s)
          phosphatidylinositol-3-phosphatase activity phosphoprotein phosphatase activity protein serine/threonine phosphatase activity protein tyrosine phosphatase activity protein binding beta-catenin binding anaphase-promoting complex binding phosphatidylinositol-3,4,5-trisphosphate 3-phosphatase activity phosphatase activity enzyme binding PDZ domain binding inositol-1,3,4,5,6-pentakisphosphate 3-phosphatase activity identical protein binding inositol-1,3,4,5-tetrakisphosphate 3-phosphatase activity phosphatidylinositol-3,4-bisphosphate 3-phosphatase activity phosphatidylinositol phosphate phosphatase activity molecular function inhibitor activity ubiquitin-specific protease binding ubiquitin ligase activator activity platelet-derived growth factor receptor binding protein kinase binding ionotropic glutamate receptor binding protein tyrosine kinase binding
          Process(es)
          protein dephosphorylation phosphatidylinositol biosynthetic process spindle assembly involved in female meiosis neuron-neuron synaptic transmission synapse assembly central nervous system development heart development learning or memory locomotory behavior positive regulation of cell proliferation negative regulation of cell proliferation regulation of signal transduction negative regulation of cell communication negative regulation of epithelial to mesenchymal transition regulation of neuron projection development negative regulation of neuron projection development cell migration dentate gyrus development central nervous system neuron axonogenesis negative regulation of signaling negative regulation of cell migration adult behavior regulation of protein stability central nervous system myelin maintenance negative regulation of peptidyl-serine phosphorylation multicellular organismal response to stress social behavior maternal behavior protein kinase B signaling locomotor rhythm regulation of cell differentiation negative regulation of osteoblast differentiation negative regulation of cell size positive regulation of transcription from RNA polymerase II promoter negative regulation of organ growth phosphatidylinositol dephosphorylation forebrain morphogenesis cell motility negative regulation of axonogenesis protein stabilization negative regulation of developmental process negative regulation of cellular component organization negative regulation of multicellular organismal process negative regulation of keratinocyte migration regulation of cell cycle negative regulation of focal adhesion assembly regulation of protein kinase B signaling negative regulation of protein kinase B signaling rhythmic synaptic transmission synapse maturation prepulse inhibition long term synaptic depression dendritic spine morphogenesis cellular response to electrical stimulus negative regulation of excitatory postsynaptic potential presynaptic membrane assembly postsynaptic density assembly positive regulation of intracellular signal transduction negative regulation of cell cycle G1/S phase transition negative regulation of wound healing, spreading of epidermal cells negative regulation of vascular smooth muscle cell proliferation positive regulation of protein ubiquitination involved in ubiquitin-dependent protein catabolic process negative regulation of G1/S transition of mitotic cell cycle positive regulation of excitatory postsynaptic potential negative regulation of cellular senescence negative regulation of synaptic vesicle clustering angiogenesis osteoblast differentiation regulation of B cell apoptotic process apoptotic process memory gene expression positive regulation of gene expression positive regulation of cardiac muscle cell apoptotic process neuron projection development negative regulation of myelination regulation of synaptic transmission, GABAergic regulation of cellular component size positive regulation of interleukin-6 production positive regulation of tumor necrosis factor production cellular response to insulin stimulus regulation of myeloid cell apoptotic process cellular response to decreased oxygen levels positive regulation of apoptotic process negative regulation of apoptotic process endothelial cell migration cellular response to leptin stimulus negative regulation of protein kinase activity by regulation of protein phosphorylation positive regulation of neuron differentiation negative regulation of cell cycle platelet-derived growth factor receptor signaling pathway regulation of axon regeneration negative regulation of axon regeneration cardiac muscle tissue development brain morphogenesis negative regulation of epithelial cell proliferation negative regulation of phagocytosis negative regulation of cardiac muscle cell proliferation male mating behavior long-term synaptic potentiation regulation of cellular localization regulation of Fc receptor mediated stimulatory signaling pathway prostate gland growth negative regulation of androgen receptor signaling pathway negative regulation of dendritic spine morphogenesis positive regulation of ERK1 and ERK2 cascade cellular response to ethanol cellular response to hypoxia inositol phosphate catabolic process negative regulation of ribosome biogenesis negative regulation of defense response to bacterium negative regulation of potassium ion transmembrane transporter activity negative regulation of dendrite extension negative regulation of TORC1 signaling cellular response to insulin-like growth factor stimulus negative regulation of receptor activity positive regulation of apoptotic signaling pathway
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-6b866bbc8c-m8c75:80/100.66.76.64:80.
          git-commit: 9d76681b284ad04e8ddcd97b79462898b32e8e5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.61.1-2026.10.99-1.0