NovaBright™ Phospha-Light™ EXP Assay Kit for SEAP (Secreted Placental Alkaline Phosphatase) Reporter Gene Detection, 2 x 96 well plates, 192 assays - FAQs
NovaBright™ Phospha-Light™ EXP Assay Kit for SEAP (Secreted Placental Alkaline Phosphatase) Reporter Gene Detection, 2 x 96 well plates, 192 assays - FAQs
View additional product information for NovaBright™ Phospha-Light™ EXP Assay Kit for SEAP (Secreted Placental Alkaline Phosphatase) Reporter Gene Detection - FAQs (N10578, N10577)
2 product FAQs found
Do you offer an alternative for SEAP Substrate Luminescence (Cat. No. A63880)?
We suggest the following products as an alternative to for SEAP Substrate Luminescence (Cat. No. A63880):
Phospha-Light SEAP Reporter Gene Assay System (Cat. Nos. T1015, T1016, T1017)
NovaBright Phospha-Light EXP Assay Kit for SEAP (Secreted Placental Alkaline Phosphatase) Reporter Gene Detection (Cat. Nos. N10577, N10578)
Both kits utilize the high-performance alkaline phosphatase substrate, CSPD, and an Emerald Enhancer.
What alkaline phosphatase substrates do you offer?