Secure-Seal™ Hybridization Chamber Gasket, 8 chambers, 9 mm diameter, 0.8 mm deep
Invitrogen™

Secure-Seal™ Hybridization Chamber Gasket, 8 chambers, 9 mm diameter, 0.8 mm deep

The Secure-Seal™ hybridization sealing systems are coverslip-seal combinations that attach to microscope slides to form microwells optimized for carrying out자세히 알아보기
Have Questions?
카탈로그 번호수량
S247321 set
카탈로그 번호 S24732
제품 가격(KRW)
276,000
온라인 행사
Ends: 15-Jul-2026
324,000
할인액 48,000 (15%)
Each
수량:
1 set
제품 가격(KRW)
276,000
온라인 행사
Ends: 15-Jul-2026
324,000
할인액 48,000 (15%)
Each
The Secure-Seal™ hybridization sealing systems are coverslip-seal combinations that attach to microscope slides to form microwells optimized for carrying out in situ hybridization procedures. These ready-to-use hybridization gaskets have a special adhesive that bonds to glass slides in seconds, creating a water-tight seal that is temperature resistant but can also be removed cleanly and easily after hybridization. Solutions are easily added or removed through dual-access ports without disrupting the specimen. The hydrophobic coverslips are nuclease free and will not trap or bind probes, allowing uniform distribution of the reagent over the specimen.
For Research Use Only. Not for use in diagnostic procedures.
사양
깊이(미터법)0.8 mm
직경(미터법)9 mm
수량1 set
배송 조건Room Temperature
제품라인Secure-Seal
유형Hybridization Chamber Gasket
Unit SizeEach
구성 및 보관
Store at room temperature.

인용 및 참조 문헌 (1)

인용 및 참조 문헌
Abstract
Mast Cell Protease 7 Promotes Angiogenesis by Degradation of Integrin Subunits.
Authors:de Souza Junior DA, Santana C, Vieira GV, Oliver C, Jamur MC
Journal:Cells
PubMed ID:31013764
'Previous studies from our laboratory have shown that during angiogenesis in vitro, rmMCP-7 (recombinant mouse mast cell protease-7) stimulates endothelial cell spreading and induces their penetration into the matrix. The ability of rmMCP-7 to induce angiogenesis in vivo was assessed in the present study using a directed in vivo angiogenesis ... More