iBind™ Flex Western Starter Kit, fluorescent detection
Invitrogen™
iBind™ Flex Western Starter Kit, fluorescent detection
The iBind Flex Western Starter Kit for fluorescent detection includes an iBind Flex Western Device , iBind Flex Cards, andRead more
Have Questions?
Catalog NumberQuantity
SLF2002S1 kit
Catalog number SLF2002S
Price (USD)
2,408.25
Special offer
Online exclusive
Ends: 04-Dec-2026
3,705.00
Save 1,296.75
1 kit
Quantity:
1 kit
Request bulk or custom format
Price (USD)
2,408.25
Special offer
Online exclusive
Ends: 04-Dec-2026
3,705.00
Save 1,296.75
1 kit
The iBind Flex Western Starter Kit for fluorescent detection includes an iBind Flex Western Device , iBind Flex Cards, and an iBind Flex Fluorescent Detection (FD) Solution Kit . The iBind Flex Western Device is a small benchtop device that automates western blot processing (immunodetection). It applies mechanical pressure to an iBind Flex Card to generate sequential lateral flow (SLF) of immunodetection solutions for the blocking, antibody binding, and washing steps. The glass fiber matrix of the iBind Flex Card allows for homogenous and consistent flow of the solutions over the membrane, increasing the antigen-antibody interaction.
The iBind Flex Western Device allows all solutions to be prepared and loaded in the device at the start of the procedure; all subsequent steps proceed automatically through SLF action—no electricity or batteries required. Blots are ready for the final detection steps in approximately 2.5 hours.
Learn more about iBind western systems ›
The iBind Flex Western Device includes three user-interchangeable well inserts to process up to one midi blot, two mini blots, or six vertically-cut strip blots when used with iBind Flex Cards, and the iBind Flex Solution Kit or iBind Flex Fluorescent Detection (FD) Solution Kit to process one mini-blot.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Detection MethodFluorescence
Membrane CompatibilityNitrocellulose, PVDF
Quantity1 kit
Shipping ConditionRoom Temperature
Throughput1 Midi Blot, 6 Vertically-Cut Strip Blots, 2 Mini Blots
Capacity1 Midi Blot, 6 Vertically-Cut Strip Blots, 2 Mini Blots
For Use With (Application)Western Blot
Product LineiBind
Running Time∼2.5 hr.
TypeStarter Kit
Unit Size1 kit
Contents & Storage
1 iBind Flex Western Device, 10 iBind Flex Cards (store at room temperature), 1 iBind Flex Fluorescent Detection (FD) Solution Kit (store at 2 to 8°C)
Frequently asked questions (FAQs)
Can I use conjugated primary antibodies with the iBind Flex Western Device?
Yes. When using a conjugated primary antibody and no secondary antibody, add the diluted primary antibody to well 1 and replace diluted secondary antibody (well 3) with 1X iBind Flex/iBind Flex FD Solution. Alternatively, for faster time to results, add the diluted primary antibody to well 1, leave wells 2 and 3 empty, and add 1X iBind Flex/ iBind Flex FD Solution to well 4. This procedure will save time (~60 min) but may result in higher background.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.
What procedure should I follow when stripping and reprobing my blot processed using the iBind Western Device or iBind Flex Western Device?
To strip and reprobe your blot processed using the iBind Western Device or iBind Flex Western Device, first strip the blot outside the system using your preferred protocol. After the stripping protocol, wash the blot in distilled water for 5 min. Reblocking is unnecessary as the iBind Solution will perform this step during reprobing. Finally, reprobe with new antibodies using the iBind Western Device or iBind Flex Western Device, starting with the membrane preparation step.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.
Can I multiplex antibodies using the iBind Western Device or iBind Flex Western Device?
Yes, for each midi blot, mini blot, or vertically cut strip, prepare appropriate primary antibodies together in a single tube of iBind FD/ iBind Flex FD Solution, Prepare appropriate secondary antibodies together in a different tube of iBind FD/ iBind Flex FD Solution. Do not mix primary and secondary antibodies prior to loading in the device. All primary and secondary antibodies should be diluted according to the recommendations outlined in the user manual or quick reference card. When detecting several targets of the same species, the same concentration of secondary antibody can be used as when detecting one target. There is no need to increase the concentration for each additional target. Load multiplexed antibodies into the device as normal.
Caution: Consider cross-reactivity of secondary antibodies when multiplexing (e.g., rabbit anti-goat IgG and goat anti-mouse IgG are likely to cross-react).
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.
Can I use the iBind Western Device or iBind Flex Western Device with Alexa Fluor Plus conjugated secondary antibodies?
Yes, both the iBind Western Device and iBind Flex Western Device work well with Alexa Fluor Plus conjugated secondary antibodies.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.
With the iBind Western Device and iBind Flex Western Device, how do I know if I am rolling the membrane firmly enough?
When firmly rolling, the iBind/iBind Flex Card will slightly dip in the membrane area, causing the top and bottom of the card to lift. See the iBind/iBind Flex user manual for additional details.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.