iBind™ Flex Fluorescent Detection (FD) Solution Kit
Invitrogen™

iBind™ Flex Fluorescent Detection (FD) Solution Kit

iBind Flex Fluorescent Detection (FD) Solution is compatible with downstream fluorescent detection (e.g., infrared dual wavelength) and serves as aRead more
Have Questions?
Catalog NumberQuantity
SLF20191 kit
Catalog number SLF2019
Price (USD)
211.65
Online exclusive
218.00
Save 6.35
1 kit
Quantity:
1 kit
Request bulk or custom format
Price (USD)
211.65
Online exclusive
218.00
Save 6.35
1 kit
iBind Flex Fluorescent Detection (FD) Solution is compatible with downstream fluorescent detection (e.g., infrared dual wavelength) and serves as a combined blocking, washing, and antibody diluent solution to process up to one midi blot, two mini blots, or six vertically cut strip blots when used with the iBind Flex Western Device and iBind Flex Cards. Each kit contains reagents sufficient for 10 midi blots or 20 mini blots.

Learn more about the iBind western systems ›

iBind Flex Western System
The iBind Flex Western System offers automated western blot processing and can accommodate various blot formats including midi, mini, and vertically cut strips. Based on Sequential Lateral Flow (SLF) technology, all blocking, antibody incubation and washing steps during immunodetection are hands-free. Compared to manual methods, the iBind Flex Western System requires less primary antibody, enables reproducible results and is compatible with downstream detection protocols.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)iBind Flex Western Device
Kit ContentsiBind™ Flex FD 5X buffer, iBind™ Flex 100X additive, iBind™ Flex FD 10% SDS
No. of Reactions10 midi blots or 20 mini blots
Quantity1 kit
Shipping ConditionApproved for shipment at Room Temperature or on Wet Ice
Detection MethodFluorescence
Membrane CompatibilityNitrocellulose, PVDF
Product LineiBind
Product TypeFlex Solution Kit
Unit Size1 kit
Contents & Storage
• iBind Flex FD 5X Buffer
• iBind Flex 100X Additive
• iBind Flex FD 10% SDS

Store at 2–8°C.

Frequently asked questions (FAQs)

Can I use conjugated primary antibodies with the iBind Flex Western Device?

Yes. When using a conjugated primary antibody and no secondary antibody, add the diluted primary antibody to well 1 and replace diluted secondary antibody (well 3) with 1X iBind Flex/iBind Flex FD Solution. Alternatively, for faster time to results, add the diluted primary antibody to well 1, leave wells 2 and 3 empty, and add 1X iBind Flex/ iBind Flex FD Solution to well 4. This procedure will save time (~60 min) but may result in higher background.

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.

What procedure should I follow when stripping and reprobing my blot processed using the iBind Western Device or iBind Flex Western Device?

To strip and reprobe your blot processed using the iBind Western Device or iBind Flex Western Device, first strip the blot outside the system using your preferred protocol. After the stripping protocol, wash the blot in distilled water for 5 min. Reblocking is unnecessary as the iBind Solution will perform this step during reprobing. Finally, reprobe with new antibodies using the iBind Western Device or iBind Flex Western Device, starting with the membrane preparation step.

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.

Can I multiplex antibodies using the iBind Western Device or iBind Flex Western Device?

Yes, for each midi blot, mini blot, or vertically cut strip, prepare appropriate primary antibodies together in a single tube of iBind FD/ iBind Flex FD Solution, Prepare appropriate secondary antibodies together in a different tube of iBind FD/ iBind Flex FD Solution. Do not mix primary and secondary antibodies prior to loading in the device. All primary and secondary antibodies should be diluted according to the recommendations outlined in the user manual or quick reference card. When detecting several targets of the same species, the same concentration of secondary antibody can be used as when detecting one target. There is no need to increase the concentration for each additional target. Load multiplexed antibodies into the device as normal.
Caution: Consider cross-reactivity of secondary antibodies when multiplexing (e.g., rabbit anti-goat IgG and goat anti-mouse IgG are likely to cross-react).

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.

Can I use the iBind Western Device or iBind Flex Western Device with Alexa Fluor Plus conjugated secondary antibodies?

Yes, both the iBind Western Device and iBind Flex Western Device work well with Alexa Fluor Plus conjugated secondary antibodies.

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.

With the iBind Western Device and iBind Flex Western Device, how do I know if I am rolling the membrane firmly enough?

When firmly rolling, the iBind/iBind Flex Card will slightly dip in the membrane area, causing the top and bottom of the card to lift. See the iBind/iBind Flex user manual for additional details.

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.