Product References
Fungal sensing by dectin-1 directs the non-pathogenic polarization of TH17 cells through balanced type I IFN responses in human DCs.
Nature immunology
Gringhuis SI,Kaptein TM,Remmerswaal EBM,Drewniak A,Wevers BA,Theelen B,D'Haens GRAM,Boekhout T,Geijtenbeek TBH
69-7179-42 was used in Flow Cytometry /test) to show that dectin-1-mediated modulation of type I IFN responses allowed TGF-β activation and non-pathogenic TH17 cell development during fungal infections in humans.
Thu Dec 01 00:00:00 EST 2022
FABP4 facilitates inflammasome activation to induce the Treg/Th17 imbalance in preeclampsia via forming a positive feedback with IL-17A.
Molecular therapy. Nucleic acids
Chang GP,Yang XL,Liu W,Lin S,Yang SL,Zhao MY
Published figure using IL-17A monoclonal antibody (Product # 69-7179-42) in Flow Cytometry
Fri Jun 04 00:00:00 EDT 2021
Small molecule allosteric inhibitors of RORγt block Th17-dependent inflammation and associated gene expression in vivo.
PloS one
Saenz SA,Local A,Carr T,Shakya A,Koul S,Hu H,Chourb L,Stedman J,Malley J,D'Agostino LA,Shanmugasundaram V,Malona J,Schwartz CE,Beebe L,Clements M,Rajaraman G,Cho J,Jiang L,Dubrovskiy A,Kreilein M,Shimanovich R,Hamann LG,Escoubet L,Ellis JM
Published figure using IL-17A monoclonal antibody (Product # 69-7179-42) in Flow Cytometry
Wed Dec 29 00:00:00 EST 2021
YY1 regulation by miR-124-3p promotes Th17 cell pathogenicity through interaction with T-bet in rheumatoid arthritis.
JCI insight
Lin J,Tang J,Lin J,He Y,Yu Z,Jiang R,Yang B,Ou Q
Published figure using IL-17A monoclonal antibody (Product # 69-7179-42) in Flow Cytometry
Mon Nov 22 00:00:00 EST 2021
Neutrophil-Derived IL-17 Promotes Ventilator-Induced Lung Injury via p38 MAPK/MCP-1 Pathway Activation.
Frontiers in immunology
Liao X,Zhang W,Dai H,Jing R,Ye M,Ge W,Pei S,Pan L
Published figure using IL-17A monoclonal antibody (Product # 69-7179-42) in Immunohistochemistry (Paraffin)
Mon Feb 14 00:00:00 EST 2022
Th17/Treg imbalance modulates rat myocardial fibrosis and heart failure by regulating LOX expression.
Acta physiologica (Oxford, England)
Lu M,Qin X,Yao J,Yang Y,Zhao M,Sun L
Published figure using IL-17A monoclonal antibody (Product # 69-7179-42) in Neutralization
Sun Nov 01 00:00:00 EDT 2020
Senolytics prevent mt-DNA-induced inflammation and promote the survival of aged organs following transplantation.
Nature communications
Iske J,Seyda M,Heinbokel T,Maenosono R,Minami K,Nian Y,Quante M,Falk CS,Azuma H,Martin F,Passos JF,Niemann CU,Tchkonia T,Kirkland JL,Elkhal A,Tullius SG
Published figure using IL-17A monoclonal antibody (Product # 69-7179-42) in Flow Cytometry
Thu Aug 27 00:00:00 EDT 2020
Developing Human Skin Contains Lymphocytes Demonstrating a Memory Signature.
Cell reports. Medicine
Dhariwala MO,Karthikeyan D,Vasquez KS,Farhat S,Weckel A,Taravati K,Leitner EG,Clancy S,Pauli M,Piper ML,Cohen JN,Ashouri JF,Lowe MM,Rosenblum MD,Scharschmidt TC
Published figure using IL-17A monoclonal antibody (Product # 69-7179-42) in Flow Cytometry
Tue Nov 17 00:00:00 EST 2020
Reciprocal regulation of Th2 and Th17 cells by PAD2-mediated citrullination.
JCI insight
Sun B,Chang HH,Salinger A,Tomita B,Bawadekar M,Holmes CL,Shelef MA,Weerapana E,Thompson PR,Ho IC
Published figure using IL-17A monoclonal antibody (Product # 69-7179-42) in ELISA
Thu Nov 14 00:00:00 EST 2019
CXCL4 is a novel inducer of human Th17 cells and correlates with IL-17 and IL-22 in psoriatic arthritis.
European journal of immunology
Affandi AJ,Silva-Cardoso SC,Garcia S,Leijten EFA,van Kempen TS,Marut W,van Roon JAG,Radstake TRDJ
Published figure using IL-17A monoclonal antibody (Product # 69-7179-42) in Flow Cytometry
Thu Mar 01 00:00:00 EST 2018
Accumulation of T-helper 22 cells, interleukin-22 and myeloid-derived suppressor cells promotes gastric cancer progression in elderly patients.
Oncology letters
Chen X,Wang Y,Wang J,Wen J,Jia X,Wang X,Zhang H
Published figure using IL-17A monoclonal antibody (Product # 69-7179-42) in Flow Cytometry
Sun Jul 01 00:00:00 EDT 2018
Cutting Edge: Selective Oral ROCK2 Inhibitor Reduces Clinical Scores in Patients with Psoriasis Vulgaris and Normalizes Skin Pathology via Concurrent Regulation of IL-17 and IL-10.
Journal of immunology (Baltimore, Md. : 1950)
Zanin-Zhorov A,Weiss JM,Trzeciak A,Chen W,Zhang J,Nyuydzefe MS,Arencibia C,Polimera S,Schueller O,Fuentes-Duculan J,Bonifacio KM,Kunjravia N,Cueto I,Soung J,Fleischmann RM,Kivitz A,Lebwohl M,Nunez M,Woodson J,Smith SL,West RF,Berger M,Krueger JG,Ryan JL,Waksal SD
Published figure using IL-17A monoclonal antibody (Product # 69-7179-42) in ELISA
Mon May 15 00:00:00 EDT 2017
Enhancing the toolbox to study IL-17A in cattle and sheep.
Veterinary research
Wattegedera SR,Corripio-Miyar Y,Pang Y,Frew D,McNeilly TN,Palarea-Albaladejo J,McInnes CJ,Hope JC,Glass EJ,Entrican G
Published figure using IL-17A monoclonal antibody (Product # 69-7179-42) in Flow Cytometry
Sat Apr 08 00:00:00 EDT 2017
Berberine exerts an anti-inflammatory role in ocular Behcet's disease.
Molecular medicine reports
Yang Y,Wang Q,Xie M,Liu P,Qi X,Liu X,Li Z
Published figure using IL-17A monoclonal antibody (Product # 69-7179-42) in Flow Cytometry
Sun Jan 01 00:00:00 EST 2017
Transcriptional signature of human pro-inflammatory TH17 cells identifies reduced IL10 gene expression in multiple sclerosis.
Nature communications
Hu D,Notarbartolo S,Croonenborghs T,Patel B,Cialic R,Yang TH,Aschenbrenner D,Andersson KM,Gattorno M,Pham M,Kivisakk P,Pierre IV,Lee Y,Kiani K,Bokarewa M,Tjon E,Pochet N,Sallusto F,Kuchroo VK,Weiner HL
Published figure using IL-17A monoclonal antibody (Product # 69-7179-42) in Flow Cytometry
Fri Nov 17 00:00:00 EST 2017
Single-cell profiling reveals GPCR heterogeneity and functional patterning during neuroinflammation.
JCI insight
Tischner D,Grimm M,Kaur H,Staudenraus D,Carvalho J,Looso M,Günther S,Wanke F,Moos S,Siller N,Breuer J,Schwab N,Zipp F,Waisman A,Kurschus FC,Offermanns S,Wettschureck N
Published figure using IL-17A monoclonal antibody (Product # 69-7179-42) in Flow Cytometry
Thu Aug 03 00:00:00 EDT 2017
CD16+ Monocyte Subset Was Enriched and Functionally Exacerbated in Driving T-Cell Activation and B-Cell Response in Systemic Lupus Erythematosus.
Frontiers in immunology
Zhu H,Hu F,Sun X,Zhang X,Zhu L,Liu X,Li X,Xu L,Shi L,Gan Y,Su Y
Published figure using IL-17A monoclonal antibody (Product # 69-7179-42) in ELISA
Wed Nov 20 00:00:00 EST 2019
CD16+ Monocyte Subset Was Enriched and Functionally Exacerbated in Driving T-Cell Activation and B-Cell Response in Systemic Lupus Erythematosus.
Frontiers in immunology
Zhu H,Hu F,Sun X,Zhang X,Zhu L,Liu X,Li X,Xu L,Shi L,Gan Y,Su Y
Published figure using IL-17A monoclonal antibody (Product # 69-7179-42) in ELISA
Wed Nov 20 00:00:00 EST 2019
Interaction among activated lymphocytes and mesenchymal cells through podoplanin is critical for a high IL-17 secretion.
Arthritis research & therapy
Noack M,Ndongo-Thiam N,Miossec P
Published figure using IL-17A monoclonal antibody (Product # 69-7179-42) in Flow Cytometry
Thu Jun 23 00:00:00 EDT 2016
Altered expression of miR-92a correlates with Th17 cell frequency in patients with primary biliary cirrhosis.
International journal of molecular medicine
Liang DY,Hou YQ,Luo LJ,Ao L
Published figure using IL-17A monoclonal antibody (Product # 69-7179-42) in Immunofluorescence
Fri Jul 01 00:00:00 EDT 2016
Altered expression of miR-92a correlates with Th17 cell frequency in patients with primary biliary cirrhosis.
International journal of molecular medicine
Liang DY,Hou YQ,Luo LJ,Ao L
Published figure using IL-17A monoclonal antibody (Product # 69-7179-42) in Immunofluorescence
Fri Jul 01 00:00:00 EDT 2016
Interleukin-25 Mediates Transcriptional Control of PD-L1 via STAT3 in Multipotent Human Mesenchymal Stromal Cells (hMSCs) to Suppress Th17 Responses.
Stem cell reports
Wang WB,Yen ML,Liu KJ,Hsu PJ,Lin MH,Chen PM,Sudhir PR,Chen CH,Chen CH,Sytwu HK,Yen BL
Published figure using IL-17A monoclonal antibody (Product # 69-7179-42) in Flow Cytometry
Tue Sep 08 00:00:00 EDT 2015
Fiber composite slices for multiplexed immunoassays.
Biomicrofluidics
Kim J,Bae S,Song S,Chung K,Kwon S
Published figure using IL-17A monoclonal antibody (Product # 69-7179-42) in Immunofluorescence
Wed Jul 01 00:00:00 EDT 2015
Fiber composite slices for multiplexed immunoassays.
Biomicrofluidics
Kim J,Bae S,Song S,Chung K,Kwon S
Published figure using IL-17A monoclonal antibody (Product # 69-7179-42) in Immunofluorescence
Wed Jul 01 00:00:00 EDT 2015
An ACT1 mutation selectively abolishes interleukin-17 responses in humans with chronic mucocutaneous candidiasis.
Immunity
Boisson B,Wang C,Pedergnana V,Wu L,Cypowyj S,Rybojad M,Belkadi A,Picard C,Abel L,Fieschi C,Puel A,Li X,Casanova JL
Published figure using IL-17A monoclonal antibody (Product # 69-7179-42) in Flow Cytometry
Thu Oct 17 00:00:00 EDT 2013
Patients with cystic fibrosis have inducible IL-17+IL-22+ memory cells in lung draining lymph nodes.
The Journal of allergy and clinical immunology
Chan YR,Chen K,Duncan SR,Lathrop KL,Latoche JD,Logar AJ,Pociask DA,Wahlberg BJ,Ray P,Ray A,Pilewski JM,Kolls JK
Published figure using IL-17A monoclonal antibody (Product # 69-7179-42) in Immunofluorescence
Mon Apr 01 00:00:00 EDT 2013
Increased frequencies of Th22 cells as well as Th17 cells in the peripheral blood of patients with ankylosing spondylitis and rheumatoid arthritis.
PloS one
Zhang L,Li YG,Li YH,Qi L,Liu XG,Yuan CZ,Hu NW,Ma DX,Li ZF,Yang Q,Li W,Li JM
Published figure using IL-17A monoclonal antibody (Product # 69-7179-42) in Flow Cytometry
Fri Jul 27 00:00:00 EDT 2012
Gain-of-function human STAT1 mutations impair IL-17 immunity and underlie chronic mucocutaneous candidiasis.
The Journal of experimental medicine
Liu L,Okada S,Kong XF,Kreins AY,Cypowyj S,Abhyankar A,Toubiana J,Itan Y,Audry M,Nitschke P,Masson C,Toth B,Flatot J,Migaud M,Chrabieh M,Kochetkov T,Bolze A,Borghesi A,Toulon A,Hiller J,Eyerich S,Eyerich K,Gulácsy V,Chernyshova L,Chernyshov V,Bondarenko A,Grimaldo RM,Blancas-Galicia L,Beas IM,Roesler J,Magdorf K,Engelhard D,Thumerelle C,Burgel PR,Hoernes M,Drexel B,Seger R,Kusuma T,Jansson AF,Sawalle-Belohradsky J,Belohradsky B,Jouanguy E,Bustamante J,Bué M,Karin N,Wildbaum G,Bodemer C,Lortholary O,Fischer A,Blanche S,Al-Muhsen S,Reichenbach J,Kobayashi M,Rosales FE,Lozano CT,Kilic SS,Oleastro M,Etzioni A,Traidl-Hoffmann C,Renner ED,Abel L,Picard C,Maródi L,Boisson-Dupuis S,Puel A,Casanova JL
Published figure using IL-17A monoclonal antibody (Product # 69-7179-42) in Flow Cytometry
Mon Aug 01 00:00:00 EDT 2011
A monoclonal antibody selection for immunohistochemical examination of lymphoid tissues from non-human primates.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society
Kap YS,van Meurs M,van Driel N,Koopman G,Melief MJ,Brok HP,Laman JD,'t Hart BA
Published figure using IL-17A monoclonal antibody (Product # 69-7179-42) in Immunohistochemistry
Tue Dec 01 00:00:00 EST 2009
Interleukin-12 is the optimum cytokine to expand human Th17 cells in vitro.
Clinical and vaccine immunology : CVI
Nady S,Ignatz-Hoover J,Shata MT
Published figure using IL-17A monoclonal antibody (Product # 69-7179-42) in ELISA
Mon Jun 01 00:00:00 EDT 2009
Interleukin-12 is the optimum cytokine to expand human Th17 cells in vitro.
Clinical and vaccine immunology : CVI
Nady S,Ignatz-Hoover J,Shata MT
Published figure using IL-17A monoclonal antibody (Product # 69-7179-42) in ELISA
Mon Jun 01 00:00:00 EDT 2009
Prolonged, granulocyte-macrophage colony-stimulating factor-dependent, neutrophil survival following rheumatoid synovial fibroblast activation by IL-17 and TNFalpha.
Arthritis research & therapy
Parsonage G,Filer A,Bik M,Hardie D,Lax S,Howlett K,Church LD,Raza K,Wong SH,Trebilcock E,Scheel-Toellner D,Salmon M,Lord JM,Buckley CD
Published figure using IL-17A monoclonal antibody (Product # 69-7179-42) in Immunofluorescence
Tue Sep 23 00:00:00 EDT 2008