Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
AXIN1
Transcript:
NM_003502.3
Assay Name:
Mutation genome location:
chr.16 287754-288262 on build GRCh38
Assay ID
Hs00150479_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Between 400-700 bp
Panther Classification:
Molecular Function -
scaffold/adaptor protein
Gene Ontology Categories:
Function(s)
Process(es)
optic placode formation
positive regulation of protein phosphorylation
apoptotic process
activation of JUN kinase activity
determination of left/right symmetry
cell death
dorsal/ventral axis specification
positive regulation of peptidyl-threonine phosphorylation
Wnt signaling pathway
forebrain anterior/posterior pattern specification
Wnt-activated signaling pathway involved in forebrain neuron fate commitment
negative regulation of Wnt signaling pathway
olfactory placode formation
cytoplasmic microtubule organization
positive regulation of protein ubiquitination
activation of protein kinase activity
positive regulation of peptidyl-serine phosphorylation
negative regulation of transcription elongation from RNA polymerase II promoter
regulation of catenin import into nucleus
proteasome-mediated ubiquitin-dependent protein catabolic process
positive regulation of GTPase activity
cellular protein complex assembly
negative regulation of fat cell differentiation
positive regulation of protein catabolic process
positive regulation of transcription, DNA-templated
positive regulation of JNK cascade
embryonic eye morphogenesis
axial mesoderm formation
positive regulation of ubiquitin-protein transferase activity
muscle cell development
canonical Wnt signaling pathway
embryonic skeletal joint morphogenesis
canonical Wnt signaling pathway involved in neural plate anterior/posterior pattern formation
cellular response to organic cyclic compound
negative regulation of canonical Wnt signaling pathway
Wnt signaling pathway involved in somitogenesis
positive regulation of canonical Wnt signaling pathway
beta-catenin destruction complex assembly
beta-catenin destruction complex disassembly
positive regulation of protein ubiquitination involved in ubiquitin-dependent protein catabolic process
signal transducer activity
GTPase activator activity
protein binding
beta-catenin binding
enzyme binding
protein kinase binding
receptor signaling complex scaffold activity
ubiquitin protein ligase binding
protein complex scaffold
identical protein binding
protein homodimerization activity
SMAD binding
armadillo repeat domain binding
I-SMAD binding
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-744bc48644-fvx7n:80/100.66.78.247:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline