Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
PTGER3
Transcript:
Assay Name:
Mutation genome location:
chr.1 71047578-71048086 on build GRCh38
Gene:
ZRANB2-AS1
Transcript:
Assay Name:
Mutation genome location:
chr.1 71047578-71048086 on build GRCh38
Assay ID
Hs00210706_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Between 400-700 bp
Panther Classification:
Molecular Function -
G-protein coupled receptor
Gene Ontology Categories:
Function(s)
Process(es)
inflammatory response
G-protein coupled receptor signaling pathway
adenylate cyclase-activating G-protein coupled receptor signaling pathway
phospholipase C-activating G-protein coupled receptor signaling pathway
positive regulation of cytosolic calcium ion concentration
female pregnancy
cell death
positive regulation of gene expression
negative regulation of norepinephrine secretion
intestine smooth muscle contraction
urinary bladder smooth muscle contraction
negative regulation of cAMP biosynthetic process
positive regulation of cAMP biosynthetic process
positive regulation of fever generation
response to estradiol
response to progesterone
response to drug
positive regulation of vasoconstriction
positive regulation of circadian sleep/wake cycle, non-REM sleep
negative regulation of insulin secretion
maternal process involved in parturition
negative regulation of gastric acid secretion
cellular response to lipopolysaccharide
cellular response to interleukin-1
negative regulation of platelet aggregation
negative regulation of potassium ion transmembrane transport
response to salt
negative regulation of calcium ion transmembrane transport
negative regulation of gastrin-induced gastric acid secretion
positive regulation of smooth muscle contraction involved in micturition
cellular response to forskolin
positive regulation of inhibitory G-protein coupled receptor phosphorylation
negative regulation of circadian sleep/wake cycle, wakefulness
positive regulation of myofibroblast contraction
positive regulation of colon smooth muscle contraction
positive regulation of gastric mucosal blood circulation
prostaglandin receptor internalization
positive regulation of neutrophil extravasation
negative regulation of forebrain neuron differentiation
prostaglandin E receptor activity
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-744bc48644-nrcn4:80/100.66.78.247:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline