Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
RARA
Transcript:
NM_001024809.3
Assay Name:
Mutation genome location:
chr.17 40355917-40356423 on build GRCh38
Assay ID
Hs00398587_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Between 400-700 bp
Panther Classification:
Molecular Function -
C4 zinc finger nuclear receptor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
ureteric bud development
neural tube closure
liver development
glandular epithelial cell development
outflow tract septum morphogenesis
growth plate cartilage development
transcription initiation from RNA polymerase II promoter
protein phosphorylation
signal transduction
germ cell development
spermatogenesis
female pregnancy
positive regulation of cell proliferation
negative regulation of cell proliferation
hippocampus development
prostate gland development
negative regulation of granulocyte differentiation
embryonic camera-type eye development
regulation of myelination
response to estradiol
response to retinoic acid
negative regulation of interferon-gamma production
negative regulation of tumor necrosis factor production
positive regulation of interleukin-13 production
positive regulation of interleukin-4 production
positive regulation of interleukin-5 production
response to vitamin A
response to cytokine
multicellular organism growth
negative regulation of apoptotic process
apoptotic cell clearance
steroid hormone mediated signaling pathway
response to ethanol
positive regulation of T-helper 2 cell differentiation
positive regulation of neuron differentiation
positive regulation of cell cycle
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
negative regulation of translational initiation
regulation of synaptic plasticity
retinoic acid receptor signaling pathway
positive regulation of binding
ventricular cardiac muscle cell differentiation
Sertoli cell fate commitment
limb development
face development
trachea cartilage development
chondroblast differentiation
negative regulation of cartilage development
cellular response to lipopolysaccharide
cellular response to retinoic acid
cellular response to estrogen stimulus
translation repressor activity, nucleic acid binding
RNA polymerase II regulatory region sequence-specific DNA binding
retinoic acid binding
transcription factor activity, sequence-specific DNA binding
steroid hormone receptor activity
retinoic acid receptor activity
transcription coactivator activity
transcription corepressor activity
receptor binding
protein binding
transcription factor binding
drug binding
zinc ion binding
enzyme binding
protein domain specific binding
chromatin DNA binding
histone deacetylase binding
protein kinase B binding
retinoic acid-responsive element binding
protein heterodimerization activity
mRNA 5'-UTR binding
protein kinase A binding
alpha-actinin binding
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline