Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
KIT
Transcript:
NM_000222.2
Assay Name:
Mutation genome location:
chr.4 54698238-54698398 on build GRCh38
Assay ID
Hs00523430_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Less than 200 bp
Panther Classification:
Molecular Function -
transmembrane signal receptor
Gene Ontology Categories:
Function(s)
Process(es)
MAPK cascade
activation of MAPK activity
ovarian follicle development
myeloid progenitor cell differentiation
lymphoid progenitor cell differentiation
immature B cell differentiation
dendritic cell cytokine production
mast cell chemotaxis
glycosphingolipid metabolic process
inflammatory response
signal transduction
spermatogenesis
spermatid development
positive regulation of cell proliferation
germ cell migration
regulation of cell shape
visual learning
male gonad development
positive regulation of gene expression
positive regulation of phospholipase C activity
regulation of phosphatidylinositol 3-kinase signaling
positive regulation of phosphatidylinositol 3-kinase signaling
peptidyl-tyrosine phosphorylation
cytokine-mediated signaling pathway
stem cell population maintenance
lamellipodium assembly
hemopoiesis
T cell differentiation
erythrocyte differentiation
melanocyte differentiation
positive regulation of cell migration
positive regulation of pseudopodium assembly
actin cytoskeleton reorganization
mast cell cytokine production
somatic stem cell population maintenance
embryonic hemopoiesis
ectopic germ cell programmed cell death
hematopoietic stem cell migration
megakaryocyte development
Fc receptor signaling pathway
Kit signaling pathway
erythropoietin-mediated signaling pathway
regulation of cell proliferation
positive regulation of tyrosine phosphorylation of Stat1 protein
positive regulation of tyrosine phosphorylation of Stat3 protein
positive regulation of tyrosine phosphorylation of Stat5 protein
negative regulation of programmed cell death
mast cell degranulation
positive regulation of MAPK cascade
pigmentation
positive regulation of GTPase activity
positive regulation of phosphatidylinositol 3-kinase activity
positive regulation of Notch signaling pathway
positive regulation of JAK-STAT cascade
protein autophosphorylation
phosphatidylinositol phosphorylation
phosphatidylinositol-mediated signaling
regulation of developmental pigmentation
somatic stem cell division
positive regulation of long-term neuronal synaptic plasticity
digestive tract development
stem cell differentiation
epithelial cell proliferation
detection of mechanical stimulus involved in sensory perception of sound
positive regulation of sequence-specific DNA binding transcription factor activity
cell chemotaxis
mast cell differentiation
mast cell proliferation
cellular response to thyroid hormone stimulus
melanocyte migration
melanocyte adhesion
positive regulation of vascular smooth muscle cell differentiation
protease binding
protein tyrosine kinase activity
transmembrane receptor protein tyrosine kinase activity
receptor signaling protein tyrosine kinase activity
stem cell factor receptor activity
Ras guanyl-nucleotide exchange factor activity
protein binding
ATP binding
cytokine binding
protein homodimerization activity
metal ion binding
phosphatidylinositol-4,5-bisphosphate 3-kinase activity
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline