Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
MTOR
Transcript:
NM_004958.3
Assay Name:
Mutation genome location:
chr.1 11114293-11114462 on build GRCh38
Assay ID
Hs00525277_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Less than 200 bp
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
protein modifying enzyme
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of protein phosphorylation
positive regulation of protein phosphorylation
positive regulation of endothelial cell proliferation
heart morphogenesis
heart valve morphogenesis
regulation of glycogen biosynthetic process
energy reserve metabolic process
'de novo' pyrimidine nucleobase biosynthetic process
DNA repair
protein phosphorylation
response to stress
cell cycle arrest
signal transduction
germ cell development
brain development
cell aging
response to nutrient
long-term memory
visual learning
post-embryonic development
negative regulation of autophagy
positive regulation of lamellipodium assembly
positive regulation of gene expression
positive regulation of myotube differentiation
positive regulation of neuron maturation
negative regulation of muscle atrophy
cell growth
macroautophagy
negative regulation of macroautophagy
phosphorylation
peptidyl-serine phosphorylation
peptidyl-threonine phosphorylation
spinal cord development
protein catabolic process
positive regulation of actin filament polymerization
T cell costimulation
negative regulation of protein ubiquitination
ruffle organization
regulation of myelination
cellular response to nutrient levels
TOR signaling
regulation of fatty acid beta-oxidation
regulation of response to food
response to insulin
regulation of actin cytoskeleton organization
social behavior
multicellular organism growth
growth
wound healing
response to cocaine
regulation of GTPase activity
response to amino acid
response to morphine
regulation of carbohydrate utilization
positive regulation of nitric oxide biosynthetic process
regulation of osteoclast differentiation
positive regulation of translation
negative regulation of cell size
regulation of protein kinase activity
positive regulation of transcription from RNA polymerase III promoter
protein autophosphorylation
positive regulation of lipid biosynthetic process
phosphatidylinositol-mediated signaling
mRNA stabilization
positive regulation of smooth muscle cell proliferation
positive regulation of oligodendrocyte differentiation
positive regulation of peptidyl-tyrosine phosphorylation
voluntary musculoskeletal movement
positive regulation of stress fiber assembly
negative regulation of NFAT protein import into nucleus
positive regulation of protein kinase B signaling
cardiac muscle cell development
cardiac muscle contraction
maternal process involved in female pregnancy
positive regulation of glial cell proliferation
positive regulation of dendritic spine development
positive regulation of cell growth involved in cardiac muscle cell development
cellular response to hypoxia
regulation of brown fat cell differentiation
regulation of membrane permeability
regulation of cellular response to heat
positive regulation of neuron death
positive regulation of transcription of nuclear large rRNA transcript from RNA polymerase I promoter
positive regulation of eating behavior
positive regulation of cholangiocyte proliferation
positive regulation of sensory perception of pain
negative regulation of cholangiocyte apoptotic process
positive regulation of granulosa cell proliferation
positive regulation of skeletal muscle hypertrophy
negative regulation of iodide transmembrane transport
RNA polymerase III type 1 promoter DNA binding
RNA polymerase III type 2 promoter DNA binding
RNA polymerase III type 3 promoter DNA binding
TFIIIC-class transcription factor binding
protein kinase activity
protein serine/threonine kinase activity
protein binding
ATP binding
kinase activity
protein kinase binding
protein domain specific binding
ribosome binding
phosphoprotein binding
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline