Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
PDPK1
Transcript:
NM_002613.4
Assay Name:
Mutation genome location:
chr.16 2586777-2587030 on build GRCh38
Assay ID
Hs00589619_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Between 200-400 bp
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
stimulatory C-type lectin receptor signaling pathway
type B pancreatic cell development
transcription, DNA-templated
regulation of transcription, DNA-templated
protein phosphorylation
negative regulation of protein kinase activity
hyperosmotic response
epidermal growth factor receptor signaling pathway
positive regulation of phospholipase activity
regulation of endothelial cell migration
negative regulation of cardiac muscle cell apoptotic process
cell migration
peptidyl-serine phosphorylation
peptidyl-threonine phosphorylation
calcium-mediated signaling
actin cytoskeleton organization
platelet activation
negative regulation of transforming growth factor beta receptor signaling pathway
T cell costimulation
activation of protein kinase B activity
cellular response to insulin stimulus
negative regulation of toll-like receptor signaling pathway
intracellular signal transduction
Fc-epsilon receptor signaling pathway
regulation of I-kappaB kinase/NF-kappaB signaling
regulation of mast cell degranulation
protein autophosphorylation
phosphatidylinositol-mediated signaling
focal adhesion assembly
T cell receptor signaling pathway
positive regulation of release of sequestered calcium ion into cytosol
cellular response to epidermal growth factor stimulus
positive regulation of establishment of protein localization to plasma membrane
extrinsic apoptotic signaling pathway
protein serine/threonine kinase activity
3-phosphoinositide-dependent protein kinase activity
insulin receptor binding
protein binding
ATP binding
phospholipase activator activity
kinase activity
protein kinase binding
phospholipase binding
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-pgj68:80/100.66.79.163:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline