Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
PSMB4
Transcript:
NM_002796.2
Assay Name:
Mutation genome location:
chr.1 151400387-151400638 on build GRCh38
Assay ID
Hs00792972_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Between 200-400 bp
Panther Classification:
Molecular Function -
protease
Gene Ontology Categories:
Function(s)
Process(es)
MAPK cascade
protein polyubiquitination
stimulatory C-type lectin receptor signaling pathway
antigen processing and presentation of exogenous peptide antigen via MHC class I, TAP-dependent
negative regulation of inflammatory response to antigenic stimulus
regulation of cellular amino acid metabolic process
viral process
anaphase-promoting complex-dependent catabolic process
tumor necrosis factor-mediated signaling pathway
NIK/NF-kappaB signaling
Fc-epsilon receptor signaling pathway
proteasome-mediated ubiquitin-dependent protein catabolic process
regulation of mRNA stability
T cell receptor signaling pathway
negative regulation of ubiquitin-protein ligase activity involved in mitotic cell cycle
positive regulation of ubiquitin-protein ligase activity involved in regulation of mitotic cell cycle transition
Wnt signaling pathway, planar cell polarity pathway
negative regulation of canonical Wnt signaling pathway
positive regulation of canonical Wnt signaling pathway
lipopolysaccharide binding
threonine-type endopeptidase activity
protein binding
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-78576c459-hhv5p:80/100.66.74.186:80.
git-commit: 65c7ea78bfc366f1196f6925435a6e3b75c9e6fa
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline