Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00366095_cn
          See other LOC105375907 CNV Assays ›
          Gene Symbol
          LOC105375907
          Assay Reference Genome
          Location

          Chr.8:77000116 on build GRCh38
          Cytoband
          8q21.13
          Assay ID Hs00366095_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          105375907

          Gene Name:

          uncharacterized LOC105375907

          Gene Aliases:

          -

          Location:

          Chr.8:76993414-77007572 on Build GRCh38

          Assay Gene Location:

          Overlaps Intron 1 - Exon 2
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          LOC105375907 XM_017014125.1 XP_016869614.1

          Target Gene Details

          Entrez Gene ID:

          5828

          Gene Name:

          peroxisomal biogenesis factor 2

          Gene Aliases:

          PAF1, PBD5A, PBD5B, PMP3, PMP35, PXMP3, RNF72, ZWS3

          Location:

          Chr.8:76980258-77001044 on Build GRCh38

          Assay Gene Location:

          Within Exon 2
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          PEX2 NM_000318.2 1 173 NP_000309.1
          NM_001079867.1 1 173 NP_001073336.1
          NM_001172086.1 NP_001165557.1
          NM_001172087.1 NP_001165558.1
          AK312997.1
          BC000661.2 AAH00661.1
          BC005375.1 AAH05375.1
          BC093043.1 AAH93043.1
          BP193912.1
          BU561273.1
          DA210382.1 1 173
          M85038.1 AAA60141.1
          M86852.1 1 88 AAC12785.1

          Back To Top

          More Information


          Set Membership:

          Intragenic Exonic Non-exonic non-DGV Variation

          Gene Ontology Categories:

          Function(s) Process(es)

          very long-chain fatty acid metabolic process
          negative regulation of transcription from RNA polymerase II promoter
          fatty acid beta-oxidation
          peroxisome organization
          protein import into peroxisome matrix
          protein destabilization
          negative regulation of fibroblast proliferation
          negative regulation of epithelial cell proliferation
          protein binding
          zinc ion binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-6b469b8bb7-5xwv5:80/100.66.76.242:80.
          git-commit: a334af76dff23450325448aedefe62379591458a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.1-2026.04.16-1.0