Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Hs02148122_cn
See other PPOX CNV Assays ›
Gene Symbol
PPOX
Assay Reference Genome
Location
Chr.1:161167505 on build GRCh38
Cytoband
1q23.3
Assay ID
Hs02148122_cn
Size
S: 360 rxns
M: 750 rxns
L: 2,900 rxns
Availability
Made To Order
Catalog #
4400291
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Target Gene Details
Entrez Gene ID:
5498
Gene Name:
protoporphyrinogen oxidase
Gene Aliases:
PPO, V290M, VP
Location:
Chr.1:161165728-161178277 on Build GRCh38
Assay Gene Location:
Within Exon 5
Gene Symbol
Transcript Accession
Exon Location
Assay Transcript Location
Protein ID
PPOX
NM_000309.3
NP_000300.1
NM_001122764.1
NP_001116236.1
XM_005245291.3
XP_005245348.2
XM_005245295.3
XP_005245352.2
XM_006711402.2
XP_006711465.2
XM_006711403.2
XP_006711466.2
XM_006711404.3
XP_006711467.2
XM_006711406.2
XP_006711469.2
XM_011509663.2
XP_011507965.1
XM_011509664.1
XP_011507966.1
XM_011509665.2
XP_011507967.1
XM_011509666.2
XP_011507968.1
XM_011509667.2
XP_011507969.1
XM_011509668.2
XP_011507970.1
XM_011509669.1
XP_011507971.1
XM_011509670.2
XP_011507972.1
XM_011509671.1
XP_011507973.1
XM_011509672.2
XP_011507974.1
XM_011509673.2
XP_011507975.1
XM_011509674.2
XP_011507976.1
XM_011509675.2
XP_011507977.1
XM_011509676.1
XP_011507978.1
XM_011509677.1
XP_011507979.1
XM_011509678.1
XP_011507980.1
XM_011509679.1
XP_011507981.1
XM_011509681.1
XP_011507983.1
XM_011509682.1
XP_011507984.1
XM_017001559.1
XP_016857048.1
XM_017001560.1
XP_016857049.1
XM_017001561.1
4
610
XP_016857050.1
XM_017001562.1
XP_016857051.1
XM_017001563.1
XP_016857052.1
XM_017001565.1
XP_016857054.1
XM_017001566.1
XP_016857055.1
XM_017001567.1
XP_016857056.1
XM_017001568.1
XP_016857057.1
XM_017001569.1
XP_016857058.1
XM_017001570.1
XP_016857059.1
XM_017001571.1
XP_016857060.1
AK094855.1
AK298912.1
AK302298.1
BC002357.2
AAH02357.1
BX384814.2
D38537.1
BAA07538.1
DC329287.1
U26446.1
AAA67690.1
Back To Top
More Information
Set Membership:
Intragenic
Exonic
non-DGV Variation
Panther Classification:
Molecular Function -
oxidase
Gene Ontology Categories:
Function(s)
Process(es)
porphyrin-containing compound biosynthetic process
protoporphyrinogen IX biosynthetic process
heme biosynthetic process
response to drug
oxidation-reduction process
oxygen-dependent protoporphyrinogen oxidase activity
flavin adenine dinucleotide binding
Back To Top
Related Products
TaqMan
®
Genotyping Master Mix
TaqPath
®
ProAmp
®
Master Mix
TaqMan
®
Universal Master Mix II, no UNG
TaqMan
®
Gene Expression Master Mix
TaqMan
®
Universal PCR Master Mix
TaqMan
®
Universal PCR Master Mix, No AmpErase UNG
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-5cvsx:80/100.66.79.163:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline