Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03175886_cn
          See other H2BC13 CNV Assays ›
          Gene Symbol
          H2BC13
          Assay Reference Genome
          Location

          Chr.6:27807908 on build GRCh38
          Cytoband
          6p22.1
          Assay ID Hs03175886_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          8340

          Gene Name:

          H2B clustered histone 13

          Gene Aliases:

          H2B/c, H2BFC, HIST1H2BL, dJ97D16.4

          Location:

          Chr.6:27807479-27807929 on Build GRCh38

          Assay Gene Location:

          Overlaps - Exon 1
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          H2BC13 NM_003519.4 NP_003510.1
          AK312114.1 1 63 BAG35050.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2025-12-01, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          esv3890803 Chr.6:27771601 - 27814760 on Build GRCh38 Loss H3C10 H2BC13 H2AC13 H2AC14
          nsv1017057 Chr.6:27777998 - 27821905 on Build GRCh38 Gain H2BC14 H3C10 H2BC13 H2AC13 H2AC14
          nsv5462086 Chr.6:27798000 - 27892000 on Build GRCh38 Duplication H3C11 H2BC14 H3C10 H4C11 H2BC13 H1-5 H3C12 H2BC15 H4C13 H4C12 H2AC15 H2AC16 H2AC13 H2AC14
          nsv5466550 Chr.6:27711907 - 27849290 on Build GRCh38 Duplication H2BC15 H4C12 H2BC14 H3C10 H2AC15 H4C11 H2BC13 H2AC13 LOC100131289 H2AC14
          nsv4132442 Chr.6:27806597 - 27814680 on Build GRCh38 Deletion H3C10 H2BC13 H2AC13 H2AC14
          nsv6398741 Chr.6:27314958 - 28461291 on Build GRCh38 Duplication OR2B2 OR2B6 ZSCAN31 ZSCAN12 H3C10 ZKSCAN4 H2BC13 H2AC17 H1-5 ZSCAN16-AS1 ZSCAN16 ZNF204P H3C12 ZKSCAN8P1 NKAPL H2BC15 LINC01012 LOC100131289 ZNF391 H2AC14 OR2B8 H3C11 PGBD1 ZNF165 ZKSCAN8 VN1R10P H2BC14 ZSCAN26 H4C11 ZSCAN12P1 ZNF184 TOB2P1 H2BC17 H4C13 ZSCAN23 H4C12 ZKSCAN3 H2AC15 H2AC16 H2AC13 ZSCAN9
          nsv6411096 Chr.6:27806501 - 27808600 on Build GRCh38 Duplication H2BC13 H2AC13
          nsv6407191 Chr.6:27684847 - 27902588 on Build GRCh38 Duplication H3C11 H2BC14 H3C10 H4C11 H2BC13 H2AC17 H1-5 H3C12 H2BC17 H2BC15 H4C13 H4C12 LINC01012 H2AC15 H2AC16 H2AC13 LOC100131289 H2AC14
          nsv830618 Chr.6:27625495 - 27821827 on Build GRCh38 Gain H2BC14 LINC01012 H3C10 H2BC13 H2AC13 LOC100131289 H2AC14
          nsv969366 Chr.6:27807410 - 27808693 on Build GRCh38 Duplication H2BC13 H2AC13
          nsv6400159 Chr.6:27753532 - 28002885 on Build GRCh38 Duplication H3C11 OR2B2 OR2B6 H2BC14 H3C10 H4C11 H2BC13 H2AC17 H1-5 H3C12 H2BC17 H2BC15 H4C13 H4C12 H2AC15 H2AC16 H2AC13 LOC100131289 H2AC14
          nsv4434899 Chr.6:27807223 - 27839222 on Build GRCh38 Duplication H2BC15 H4C12 H2BC14 H3C10 H2AC15 H4C11 H2BC13 H2AC13 H2AC14

          Back To Top

          More Information


          Additional Information:

          Set Membership:

          Intragenic Non-exonic DGV Variation Copy Number Validated

          Gene Ontology Categories:

          Function(s) Process(es)

          innate immune response in mucosa
          chromatin organization
          nucleosome assembly
          antibacterial humoral response
          antimicrobial humoral immune response mediated by antimicrobial peptide
          DNA binding
          protein binding
          structural constituent of chromatin
          protein heterodimerization activity

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-frpqk:80/100.66.72.150:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch