Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04047934_cn
          See other LAMA5 CNV Assays ›
          Gene Symbol
          LAMA5
          Assay Reference Genome
          Location

          Chr.20:62337522 on build GRCh38
          Cytoband
          20q13.33
          Assay ID Hs04047934_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          3911

          Gene Name:

          laminin subunit alpha 5

          Gene Aliases:

          -

          Location:

          Chr.20:62309060-62367318 on Build GRCh38

          Assay Gene Location:

          Within Intron 16
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          LAMA5 NM_005560.4 NP_005551.3
          XM_006723796.3 XP_006723859.1
          XM_006723798.3 XP_006723861.1
          XM_011528818.2 XP_011527120.1
          XM_011528819.2 XP_011527121.1
          AB067494.1 BAB67800.1
          AF443072.1 AAM12527.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2016-05-15, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          esv2422398 Chr.20:62207833 - 62674040 on Build GRCh38 Duplication LAMA5 ADRM1 MIR133A2 SLCO4A1-AS1 HRH3 MIR1-1HG CABLES2 LOC105369209 LAMA5-AS1 RPS21 MIR1-1 OSBPL2 SLCO4A1 RBBP8NL MIR4758 GATA5 C20orf166-AS1
          nsv459065 Chr.20:62332390 - 62812195 on Build GRCh38 Gain LAMA5 LOC107987285 LOC107985428 MIR133A2 SLCO4A1-AS1 MIR1-1HG OGFR-AS1 NTSR1 CABLES2 LAMA5-AS1 RPS21 MIR1-1 SLCO4A1 OGFR RBBP8NL MIR4758 GATA5 LINC00659 MRGBP C20orf166-AS1
          nsv953301 Chr.20:62305045 - 62338544 on Build GRCh38 Deletion LAMA5 ADRM1 MIR4758
          nsv520024 Chr.20:62312525 - 62429086 on Build GRCh38 Loss CABLES2 LAMA5 LAMA5-AS1 RPS21 RBBP8NL MIR4758
          nsv518296 Chr.20:62203245 - 62415019 on Build GRCh38 Loss CABLES2 LAMA5 ADRM1 MTG2 LOC105369209 LAMA5-AS1 RPS21 OSBPL2 RBBP8NL HRH3 MIR4758
          dgv518n27 Chr.20:62315508 - 62412954 on Build GRCh38 Loss CABLES2 LAMA5 LAMA5-AS1 RPS21 RBBP8NL MIR4758
          nsv470556 Chr.20:62212004 - 62577283 on Build GRCh38 Loss LAMA5 ADRM1 MIR133A2 HRH3 MIR1-1HG CABLES2 LOC105369209 LAMA5-AS1 RPS21 MIR1-1 OSBPL2 RBBP8NL MIR4758 GATA5 C20orf166-AS1
          nsv834024 Chr.20:62259914 - 62455522 on Build GRCh38 Gain CABLES2 LAMA5 ADRM1 LAMA5-AS1 RPS21 OSBPL2 RBBP8NL MIR4758

          Back To Top

          More Information


          Set Membership:

          Intragenic Intronic Non-exonic DGV Variation

          Panther Classification:

          Molecular Function -

          extracellular matrix protein

          Gene Ontology Categories:

          Function(s) Process(es)

          angiogenesis
          branching involved in ureteric bud morphogenesis
          morphogenesis of a polarized epithelium
          neural crest cell migration
          hair follicle development
          cytoskeleton organization
          integrin-mediated signaling pathway
          muscle organ development
          cell recognition
          cell proliferation
          embryo development
          morphogenesis of embryonic epithelium
          cell migration
          cell differentiation
          regulation of cell adhesion
          extracellular matrix organization
          lung development
          regulation of cell migration
          substrate adhesion-dependent cell spreading
          regulation of cell proliferation
          cilium assembly
          odontogenesis of dentin-containing tooth
          endothelial cell differentiation
          regulation of embryonic development
          focal adhesion assembly
          branching involved in salivary gland morphogenesis
          establishment of protein localization to plasma membrane
          integrin binding
          structural molecule activity

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline