Search
Search

Entrez Gene ID: | 102465134 |
Gene Name: | microRNA 6126 |
Gene Aliases: |
hsa-mir-6126 |
Location: |
Chr.16:3485381-3485469 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MIR6126 | NR_106741.1 |
Entrez Gene ID: | 79903 |
Gene Name: | N(alpha)-acetyltransferase 60, NatF catalytic subunit |
Gene Aliases: |
HAT4, NAT15 |
Location: |
Chr.16:3443611-3486963 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NAA60 | NM_001083600.2 | NP_001077069.1 | ||
| NM_001083601.2 | NP_001077070.1 | |||
| NM_001317093.1 | NP_001304022.1 | |||
| NM_001317094.1 | 4 | 1199 | NP_001304023.1 | |
| NM_001317095.1 | NP_001304024.1 | |||
| NM_001317096.1 | NP_001304025.1 | |||
| NM_001317097.1 | NP_001304026.1 | |||
| NM_001317098.1 | NP_001304027.1 | |||
| NM_024845.3 | NP_079121.1 | |||
| AK024216.1 | BAB14853.1 | |||
| AK092005.1 | ||||
| AK095819.1 | ||||
| AK130792.1 | ||||
| AK297219.1 | ||||
| AK298566.1 | ||||
| AK302361.1 | ||||
| AK311524.1 | 4 | 1106 | ||
| AL833594.1 | ||||
| AY358543.1 | AAQ88907.1 | |||
| BC011267.1 | AAH11267.1 | |||
| BG472911.1 | ||||
| BG822206.1 | ||||
| BU501490.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv952908 | Chr.16:3462801 - 3499000 on Build GRCh38 | Deletion |
|
Set Membership: |
|