Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00432579_cn
          See other KMT2C CNV Assays ›
          Gene Symbol
          KMT2C
          Assay Reference Genome
          Location

          Chr.7:152171285 on build GRCh38
          Cytoband
          7q36.1
          Assay ID Hs00432579_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          58508

          Gene Name:

          lysine methyltransferase 2C

          Gene Aliases:

          HALR, MLL3

          Location:

          Chr.7:152134925-152436642 on Build GRCh38

          Assay Gene Location:

          Overlaps Intron 45 - Exon 45
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          KMT2C NM_170606.2 NP_733751.2
          XM_005250025.4 XP_005250082.1
          XM_005250026.3 XP_005250083.1
          XM_005250027.4 XP_005250084.1
          XM_005250028.4 XP_005250085.1
          XM_005250031.4 XP_005250088.1
          XM_006716077.3 XP_006716140.1
          XM_006716078.3 XP_006716141.1
          XM_006716079.3 XP_006716142.1
          XM_011516450.2 XP_011514752.1
          XM_011516451.2 XP_011514753.1
          XM_011516452.2 XP_011514754.1
          XM_011516453.2 XP_011514755.1
          XM_011516454.2 XP_011514756.1
          XM_011516456.2 XP_011514758.1
          XM_017012480.1 XP_016867969.1
          XM_017012481.1 XP_016867970.1
          XM_017012482.1 XP_016867971.1
          XM_017012483.1 XP_016867972.1
          XM_017012484.1 XP_016867973.1
          XM_017012485.1 XP_016867974.1
          XM_017012486.1 XP_016867975.1
          XM_017012487.1 XP_016867976.1
          XM_017012488.1 XP_016867977.1
          XM_017012489.1 XP_016867978.1
          XM_017012490.1 XP_016867979.1
          AB040939.2 BAA96030.2
          AF264750.2 AAF74766.2
          AK025553.1
          AL832387.1
          AY024361.1 AAK00583.1
          BC151837.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2016-05-15, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          nsv609045 Chr.7:152094085 - 152201919 on Build GRCh38 Gain LOC731075 YBX1P4 KMT2C GALNT11
          nsv1024517 Chr.7:152123736 - 152424864 on Build GRCh38 Gain LOC731075 YBX1P4 KMT2C
          nsv1021676 Chr.7:152110676 - 152228495 on Build GRCh38 Gain LOC731075 YBX1P4 KMT2C GALNT11
          nsv1029778 Chr.7:151966767 - 152375920 on Build GRCh38 Gain LOC731075 YBX1P4 KMT2C GALNTL5 GALNT11
          esv3576482 Chr.7:152058842 - 152259368 on Build GRCh38 Gain LOC731075 YBX1P4 KMT2C GALNT11
          nsv831191 Chr.7:152076503 - 152260357 on Build GRCh38 Gain LOC731075 YBX1P4 KMT2C GALNT11
          nsv436540 Chr.7:152168438 - 152175074 on Build GRCh38 Deletion KMT2C
          nsv1020487 Chr.7:151946824 - 152195828 on Build GRCh38 Gain LOC731075 YBX1P4 KMT2C GALNTL5 GALNT11
          dgv6758n100 Chr.7:152070390 - 152305071 on Build GRCh38 Gain LOC731075 YBX1P4 KMT2C GALNT11
          dgv1338e214 Chr.7:152116842 - 152232964 on Build GRCh38 Gain LOC731075 YBX1P4 KMT2C GALNT11

          Back To Top

          More Information


          Set Membership:

          Intragenic Non-exonic DGV Variation

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          histone H3-K4 methylation
          DNA binding
          protein binding
          zinc ion binding
          transferase activity, transferring acyl groups
          histone-lysine N-methyltransferase activity
          histone methyltransferase activity (H3-K4 specific)
          poly(A) RNA binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-74d7b4b4d9-ncdd4:80/100.66.78.131:80.
          git-commit: 7aafa54bd143a040505a257d2a3ebda4095a617f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.56.0-Offline