Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00962004_cn
          See other KCND1 CNV Assays ›
          Gene Symbol
          KCND1
          Assay Reference Genome
          Location

          Chr.X:48969900 on build GRCh38
          Cytoband
          Xp11.23
          Assay ID Hs00962004_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          3750

          Gene Name:

          potassium voltage-gated channel subfamily D member 1

          Gene Aliases:

          KV4.1

          Location:

          Chr.X:48960983-48972099 on Build GRCh38

          Assay Gene Location:

          Within Exon 1
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          KCND1 NM_004979.5 1 1945 NP_004970.3
          XM_011543910.2 2 853 XP_011542212.1
          XM_017029508.1 2 684 XP_016884997.1
          XM_017029509.1 2 681 XP_016884998.1
          XM_017029510.1 1 1795 XP_016884999.1
          AB021865.1 1 1671 BAA96454.1
          AF166003.1 1 422 AAF65516.1
          AJ005898.1 1 372 CAA06755.1
          AK315092.1 1 856
          BC045659.1 1 1945 AAH45659.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2016-05-15, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          nsv528292 Chr.X:48433286 - 49274368 on Build GRCh38 Gain GPKOW CACNA1F TIMM17B RBM3 KCND1 GATA1 PIM2 CCDC120 WDR13 PRICKLE3 EBP TFE3 WDR45 PLP2 SYP PQBP1 PPP1R3F SUV39H1 SLC35A2 PCSK1N FOXP3 FTSJ1 GLOD5 PRAF2 OTUD5 LOC101927635 SYP-AS1 WAS SLC38A5 TBC1D25 GRIPAP1 MAGIX CCDC22 HDAC6 ERAS PORCN
          esv32909 Chr.X:48003374 - 49285646 on Build GRCh38 Gain+Loss GPKOW CACNA1F SSX4 TIMM17B RBM3 KCND1 GATA1 PIM2 ZNF630 CCDC120 WDR13 PRICKLE3 EBP SSX1 TFE3 WDR45 PLP2 SYP PQBP1 PPP1R3F SUV39H1 SLC35A2 PCSK1N SSX4B FOXP3 FTSJ1 GLOD5 SSX6 PRAF2 OTUD5 SSX9 LOC101927635 SPACA5 SYP-AS1 WAS SLC38A5 SSX3 TBC1D25 GRIPAP1 MAGIX SSX5 CCDC22 SPACA5B ZNF630-AS1 ZNF182 HDAC6 ERAS PORCN

          Back To Top

          More Information


          Set Membership:

          Intragenic Exonic DGV Variation

          Panther Classification:

          Molecular Function -

          voltage-gated ion channel

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of ion transmembrane transport
          protein homooligomerization
          cardiac conduction
          potassium ion transmembrane transport
          A-type (transient outward) potassium channel activity
          delayed rectifier potassium channel activity
          metal ion binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-6479c7f679-rv8h9:80/100.66.78.195:80.
          git-commit: b70c0dceb888abc3b0dcebb5bc3a5b8753e7e816
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.50.0-Offline