Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs02914877_cn
          See other CHRM1 CNV Assays ›
          Gene Symbol
          CHRM1
          Assay Reference Genome
          Location

          Chr.11:62910011 on build GRCh38
          Cytoband
          Assay ID Hs02914877_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          1128

          Gene Name:

          cholinergic receptor muscarinic 1

          Gene Aliases:

          HM1, M1, M1R

          Location:

          Chr.11:62908679-62921878 on Build GRCh38

          Assay Gene Location:

          Within Exon 3
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          CHRM1 NM_000738.3 2 1314 NP_000729.2
          XM_011544742.3 2 1209 XP_011543044.1
          AF385587.1 1 1100 AAK68112.1
          AF498915.1 1 1090 AAM18938.1
          AK312374.1 2 1251 BAG35292.1
          BC007740.2 2 1212 AAH07740.1
          BC022984.1 2 1491 AAH22984.1
          BT007166.1 1 1090 AAP35830.1

          Target Gene Details

          Entrez Gene ID:

          139281663

          Gene Name:

          CHRM1 antisense RNA 1

          Gene Aliases:

          -

          Location:

          Chr.11:62909407-62918361 on Build GRCh38

          Assay Gene Location:

          Within Intron 1
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          CHRM1-AS1 NR_199052.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2025-12-01, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          esv34011 Chr.11:62885307 - 63226230 on Build GRCh38 Loss SLC22A6 SLC22A24 SLC22A25 CHRM1-AS1 SLC22A8 CHRM1 SLC3A2

          Back To Top

          More Information


          Set Membership:

          Intragenic Exonic Intronic Non-exonic DGV Variation

          Gene Ontology Categories:

          Function(s) Process(es)

          signal transduction
          G-protein coupled receptor signaling pathway
          G-protein coupled receptor signaling pathway, coupled to cyclic nucleotide second messenger
          adenylate cyclase-inhibiting G-protein coupled acetylcholine receptor signaling pathway
          phospholipase C-activating G-protein coupled receptor signaling pathway
          phospholipase C-activating G-protein coupled acetylcholine receptor signaling pathway
          G-protein coupled acetylcholine receptor signaling pathway
          chemical synaptic transmission
          neuromuscular synaptic transmission
          nervous system development
          regulation of locomotion
          positive regulation of ion transport
          saliva secretion
          cognition
          regulation of postsynaptic membrane potential
          regulation of glial cell proliferation
          positive regulation of intracellular protein transport
          acetylcholine receptor signaling pathway
          postsynaptic modulation of chemical synaptic transmission
          phosphatidylinositol phospholipase C activity
          G-protein coupled receptor activity
          protein binding
          G-protein coupled acetylcholine receptor activity

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline