Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03395816_cn
          See other ACP1 CNV Assays ›
          Gene Symbol
          ACP1
          Assay Reference Genome
          Location

          Chr.2:273406 on build GRCh38
          Cytoband
          Assay ID Hs03395816_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          52

          Gene Name:

          acid phosphatase 1

          Gene Aliases:

          HAAP, LMW-PTP, LMWPTP

          Location:

          Chr.2:264947-278283 on Build GRCh38

          Assay Gene Location:

          Within Intron 3
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          ACP1 NM_004300.4 NP_004291.1
          NM_007099.4 NP_009030.1
          NR_024080.2
          AB209838.1 BAD93075.1
          AK289934.1 BAF82623.1
          AK291861.1 BAF84550.1
          BC007422.2 AAH07422.1
          BC106011.1 AAI06012.1
          BI460080.1
          BJ992018.1
          BT007136.1 AAP35800.1
          DB443811.1
          M87545.1
          S62884.1 AAB27085.1
          Y16846.1 CAA76416.1

          Target Gene Details

          Entrez Gene ID:

          7840

          Gene Name:

          ALMS1 centrosome and basal body associated protein

          Gene Aliases:

          ALSS

          Location:

          Chr.2:55253-279417 on Build GRCh38

          Assay Gene Location:

          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          ALMS1

          Target Gene Details

          Entrez Gene ID:

          728378

          Gene Name:

          POTE ankyrin domain family member F

          Gene Aliases:

          A26C1B, ACTB, POTE2alpha, POTEACTIN

          Location:

          Chr.2:241401-305918 on Build GRCh38

          Assay Gene Location:

          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          POTEF

          Target Gene Details

          Entrez Gene ID:

          54884

          Gene Name:

          retinol saturase

          Gene Aliases:

          -

          Location:

          Chr.2:268195-280766 on Build GRCh38

          Assay Gene Location:

          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          RETSAT

          Target Gene Details

          Entrez Gene ID:

          6295

          Gene Name:

          S-antigen visual arrestin

          Gene Aliases:

          RP47, RP96, S-AG

          Location:

          Chr.2:253154-292393 on Build GRCh38

          Assay Gene Location:

          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          SAG

          Target Gene Details

          Entrez Gene ID:

          8604

          Gene Name:

          solute carrier family 25 member 12

          Gene Aliases:

          AGC1, ARALAR, DEE39, EIEE39

          Location:

          Chr.2:213578-324837 on Build GRCh38

          Assay Gene Location:

          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          SLC25A12

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2025-12-01, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          esv3589598 Chr.2:250064 - 298533 on Build GRCh38 Gain ALMS1 IGKV3-20 SLC25A12 SH3YL1 POTEF LOC100506076 TGOLN2 ELMOD3 IGKV2-24 NDUFS1 EEF1B2 ACP1 SNORD51 IGK LOC105373346 ALKAL2 SNORA41 TRIM51JP SAG RETSAT CMKLR2
          esv3589599 Chr.2:251988 - 283726 on Build GRCh38 Gain ALMS1 SLC25A12 SH3YL1 POTEF LOC100506076 TGOLN2 ELMOD3 NDUFS1 EEF1B2 ACP1 SNORD51 IGK ALKAL2 SNORA41 TRIM51JP SAG RETSAT
          dgv3698n100 Chr.2:265970 - 313392 on Build GRCh38 Gain ALMS1 SLC25A12 LINC03052 POTEF LOC100506076 ELMOD3 IGKV2-24 NDUFS1 EEF1B2 ACP1 MED15P9 SNORD51 IGK LOC105373346 DGKD ALKAL2 SNORA41 ANKRD33BP1 NAT8 SAG RETSAT CMKLR2-AS CMKLR2 IGKV2-26
          nsv833159 Chr.2:158491 - 327294 on Build GRCh38 Loss TCF7L1 ALMS1 INO80D-AS1 SCARNA6 CCDC74B LINC01865 LINC03052 LOC100506076 ELMOD3 IGKV2-24 LOC124905368 MED15P9 LINC01881 DGKD ANKRD33BP1 NDUFA10 GCSHP3 SLC25A12 NDUFS1 ANKRD36 TRIM64FP TRIM51JP NAT8 SAG RETSAT IGKV3-20 TPO TRIM43 SH3YL1 FAHD2A EEF1B2 SNORD51 LOC105373346 ALMS1P1 FAR2P1 CMKLR2-AS SCARNA5 CMKLR2 ATG16L1 POTEF TGOLN2 TDRD15 DYNC1I2 INO80D ACP1 IGK ALKAL2 SNORA41 LALTOP IGKV2-26
          nsv4052979 Chr.2:10001 - 590000 on Build GRCh38 Deletion G6PC2 SCARNA6 CCDC74B LINC01822 LINC03052 MIR1245A PLAC9P1 ELMOD3 LOC101926959 LOC124905368 LINC01881 LINC01238 NEU4 NDUFA10 IGKV2-29 INPP5D RAB6C-AS1 ADI1 PDCD1 C2orf92 ANKRD36B NAT8 RETSAT COL3A1 LINC03100 GPAT2 LINC01874 IGKV3-20 TPO MIR3606 HAT1 SMPD4 NAT8B LOC100287387 SNORD51 LOC105373346 ALMS1P1 TRIM43B CMKLR2-AS LINC02572 LINC03124 SCARNA5 TCF7L1-IT1 CMKLR2 LINC01115 MZT2B ATG16L1 TRAPPC12 LINC01237 POTEF FAM240C TGOLN2 TDRD15 ZDBF2 PROM2 ACP1 IGK ALKAL2 SNORA41 LALTOP LINC01880 GPAT2P2 LOC100507443 LOC100996637 CRYGC TCF7L1 ALMS1 INO80D-AS1 LINC01865 APPAT COX5B APOB LOC100506076 IGKV2-24 NOSTRIN MED15P9 KIF5C METAP1D DGKD ACTR1B ANKRD33BP1 GAL3ST2 TWIST2 MIR4784 FAM110C CRYGD GCSHP3 TUBA3E SLC25A12 SNTG2 TRAPPC12-AS1 RAB6C KCNIP3 IGKV1-5 NOC2LP1 NDUFS1 ANKRD36 LINC01856 RTP5 TRIM64FP TRIM51JP SAG LOC124905962 TPRKB LOC285097 LOC105373352 LOC102724579 TRIM43 FAHD2B SH3YL1 DUSP11 FAHD2A MIR1245B EEF1B2 FAR2P1 LOC150935 ABCB11 ALMS1-IT1 DYNC1I2 DLX1 INO80D LINC01875 CDRT15P3 SPC25 C2orf78 IGKV2-26
          nsv6316255 Chr.2:27263 - 377948 on Build GRCh38 Duplication G6PC2 SCARNA6 CCDC74B LINC01822 LINC03052 MIR1245A PLAC9P1 ELMOD3 LOC124905368 LINC01881 NEU4 NDUFA10 IGKV2-29 INPP5D RAB6C-AS1 ADI1 PDCD1 NAT8 RETSAT COL3A1 LINC03100 GPAT2 IGKV3-20 TPO MIR3606 HAT1 SMPD4 NAT8B LOC100287387 SNORD51 LOC105373346 ALMS1P1 TRIM43B CMKLR2-AS LINC02572 LINC03124 SCARNA5 TCF7L1-IT1 CMKLR2 LINC01115 MZT2B ATG16L1 TRAPPC12 LINC01237 POTEF FAM240C TGOLN2 TDRD15 PROM2 ACP1 IGK ALKAL2 SNORA41 LALTOP LINC01880 GPAT2P2 LOC100507443 CRYGC TCF7L1 ALMS1 INO80D-AS1 LINC01865 APOB LOC100506076 IGKV2-24 NOSTRIN MED15P9 KIF5C DGKD ANKRD33BP1 TWIST2 MIR4784 FAM110C CRYGD GCSHP3 TUBA3E SLC25A12 SNTG2 TRAPPC12-AS1 RAB6C KCNIP3 IGKV1-5 NDUFS1 ANKRD36 LINC01856 RTP5 TRIM64FP TRIM51JP SAG LOC285097 LOC102724579 TRIM43 FAHD2B SH3YL1 FAHD2A MIR1245B EEF1B2 FAR2P1 LOC150935 ABCB11 ALMS1-IT1 DYNC1I2 INO80D CDRT15P3 SPC25 IGKV2-26
          nsv6321644 Chr.2:214401 - 293900 on Build GRCh38 Duplication TCF7L1 ALMS1 SCARNA6 IGKV3-20 TPO TRIM43 SH3YL1 LOC100506076 ELMOD3 IGKV2-24 EEF1B2 SNORD51 LOC105373346 FAR2P1 SCARNA5 CMKLR2 GCSHP3 SLC25A12 ATG16L1 POTEF TGOLN2 NDUFS1 ANKRD36 ACP1 IGK ALKAL2 SNORA41 TRIM51JP SAG RETSAT
          nsv6321692 Chr.2:264180 - 313687 on Build GRCh38 Duplication ALMS1 SH3YL1 LINC03052 LOC100506076 ELMOD3 IGKV2-24 EEF1B2 MED15P9 SNORD51 LOC105373346 DGKD ANKRD33BP1 CMKLR2-AS CMKLR2 SLC25A12 POTEF NDUFS1 ACP1 IGK ALKAL2 SNORA41 NAT8 SAG RETSAT IGKV2-26
          nsv4059558 Chr.2:56853 - 309597 on Build GRCh38 Duplication TCF7L1 ALMS1 INO80D-AS1 G6PC2 SCARNA6 APOB LOC100506076 MIR1245A PLAC9P1 ELMOD3 IGKV2-24 LOC124905368 MED15P9 KIF5C LINC01881 DGKD TWIST2 NDUFA10 INPP5D CRYGD RAB6C-AS1 GCSHP3 SLC25A12 ADI1 PDCD1 SNTG2 TRAPPC12-AS1 RAB6C KCNIP3 NDUFS1 ANKRD36 LINC01856 RTP5 TRIM51JP SAG RETSAT COL3A1 LINC03100 GPAT2 LOC102724579 IGKV3-20 TPO MIR3606 TRIM43 FAHD2B SH3YL1 FAHD2A MIR1245B EEF1B2 LOC100287387 SNORD51 LOC105373346 FAR2P1 LINC03124 SCARNA5 TCF7L1-IT1 LOC150935 CMKLR2 LINC01115 ABCB11 ALMS1-IT1 ATG16L1 TRAPPC12 LINC01237 POTEF FAM240C TGOLN2 TDRD15 DYNC1I2 INO80D ACP1 IGK ALKAL2 SNORA41 CDRT15P3 SPC25 LALTOP LINC01880 GPAT2P2 LOC100507443 CRYGC IGKV2-26
          esv2759016 Chr.2:167949 - 929902 on Build GRCh38 Gain TCF7L1 ALMS1 INO80D-AS1 SCARNA6 CCDC74B LINC01865 APPAT COX5B LINC03052 LOC100506076 ELMOD3 LOC101926959 IGKV2-24 LOC124905368 MED15P9 METAP1D DGKD ACTR1B ANKRD33BP1 MIR4784 TMEM18-DT NDUFA10 IGKV2-29 GCSHP3 TUBA3E SLC25A12 C2orf92 NOC2LP1 NDUFS1 ANKRD36 LOC105373480 TRIM64FP ANKRD36B TRIM51JP NAT8 SAG RETSAT LOC124905962 TPRKB LINC01874 LOC105373352 IGKV3-20 TPO TRIM43 HAT1 SMPD4 SH3YL1 DUSP11 NAT8B FAHD2A TMEM18 EEF1B2 SNORD51 LOC105373346 ALMS1P1 TRIM43B FAR2P1 CMKLR2-AS SCARNA5 CMKLR2 LINC01115 LINC01939 MZT2B ATG16L1 POTEF TMEM131 TGOLN2 TDRD15 ZAP70 DYNC1I2 ZDBF2 DLX1 INO80D ACP1 LINC01875 IGK ALKAL2 SNORA41 LALTOP C2orf78 LOC100996637 IGKV2-26
          nsv5442514 Chr.2:252077 - 294598 on Build GRCh38 Duplication ALMS1 SLC25A12 SH3YL1 POTEF LOC100506076 TGOLN2 ELMOD3 IGKV2-24 NDUFS1 EEF1B2 ACP1 SNORD51 IGK LOC105373346 ALKAL2 SNORA41 TRIM51JP SAG RETSAT CMKLR2
          dgv3779n223 Chr.2:197016 - 763147 on Build GRCh38 Duplication TCF7L1 ALMS1 SCARNA6 CCDC74B LINC01865 APPAT COX5B LINC03052 LOC100506076 ELMOD3 LOC101926959 IGKV2-24 MED15P9 METAP1D DGKD ACTR1B ANKRD33BP1 MIR4784 TMEM18-DT NDUFA10 IGKV2-29 GCSHP3 TUBA3E SLC25A12 C2orf92 NOC2LP1 NDUFS1 ANKRD36 LOC105373480 TRIM64FP ANKRD36B TRIM51JP NAT8 SAG RETSAT LOC124905962 TPRKB LINC01874 LOC105373352 IGKV3-20 TPO TRIM43 HAT1 SMPD4 SH3YL1 DUSP11 NAT8B TMEM18 EEF1B2 SNORD51 LOC105373346 ALMS1P1 TRIM43B FAR2P1 CMKLR2-AS SCARNA5 CMKLR2 MZT2B ATG16L1 POTEF TMEM131 TGOLN2 TDRD15 ZAP70 ZDBF2 DLX1 ACP1 LINC01875 IGK ALKAL2 SNORA41 C2orf78 LOC100996637 IGKV2-26

          Back To Top

          More Information


          Set Membership:

          Intragenic Intronic Non-exonic DGV Variation

          Gene Ontology Categories:

          Function(s) Process(es)

          chemical synaptic transmission
          acid phosphatase activity
          phosphoprotein phosphatase activity
          protein tyrosine phosphatase activity
          non-membrane spanning protein tyrosine phosphatase activity
          protein binding
          hydrolase activity
          phosphatase activity
          SH3 domain binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline