Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03912676_cn
          See other TNFAIP8L3 CNV Assays ›
          Gene Symbol
          TNFAIP8L3
          Assay Reference Genome
          Location

          Chr.15:51059834 on build GRCh38
          Cytoband
          15q21.2
          Assay ID Hs03912676_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          388121

          Gene Name:

          TNF alpha induced protein 8 like 3

          Gene Aliases:

          TIPE3

          Location:

          Chr.15:51056596-51105276 on Build GRCh38

          Assay Gene Location:

          Within Intron 5
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          TNFAIP8L3 NM_001311175.1 NP_001298104.1
          NM_207381.3 NP_997264.2
          XM_011521545.2 XP_011519847.1
          XM_017022168.1 XP_016877657.1
          XM_017022169.1 XP_016877658.1
          AK123281.1 BAC85572.1
          AY517501.1 AAS76642.1
          BC127701.1
          BC127702.1
          HY052681.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2016-05-15, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          esv3636460 Chr.15:51022337 - 51086008 on Build GRCh38 Gain TNFAIP8L3 PIRC66
          esv3636461 Chr.15:51022583 - 51482654 on Build GRCh38 Gain MIR4713 MIR7973-1 DMXL2 GLDN CYP19A1 MIR7973-2 TNFAIP8L3 PIRC66
          nsv471243 Chr.15:51053288 - 51725899 on Build GRCh38 Gain MIR4713 SCG3 MIR7973-1 LYSMD2 DMXL2 GLDN CYP19A1 MIR7973-2 TNFAIP8L3 PIRC66
          nsv1047443 Chr.15:50452584 - 51354826 on Build GRCh38 Gain MIR4713 MIR7973-1 GLDN USP50 CYP19A1 TRPM7 AP4E1 USP8 DCAF13P3 TNFAIP8L3 SPPL2A MIR7973-2 PIRC66
          esv3636459 Chr.15:51017650 - 51162500 on Build GRCh38 Gain TNFAIP8L3 PIRC66
          esv34532 Chr.15:50983931 - 51460659 on Build GRCh38 Gain MIR4713 MIR7973-1 DMXL2 GLDN CYP19A1 AP4E1 MIR7973-2 TNFAIP8L3 PIRC66
          esv2760031 Chr.15:50990060 - 51538142 on Build GRCh38 Gain MIR4713 MIR7973-1 DMXL2 GLDN CYP19A1 AP4E1 MIR7973-2 TNFAIP8L3 PIRC66
          nsv1038778 Chr.15:50904444 - 51064531 on Build GRCh38 Loss AP4E1 DCAF13P3 TNFAIP8L3 PIRC66
          dgv4526n54 Chr.15:50582767 - 51475068 on Build GRCh38 Gain MIR4713 MIR7973-1 DMXL2 GLDN CYP19A1 TRPM7 AP4E1 SPPL2A MIR7973-2 DCAF13P3 TNFAIP8L3 PIRC66

          Back To Top

          More Information


          Set Membership:

          Intragenic Intronic Non-exonic DGV Variation

          Gene Ontology Categories:

          Function(s) Process(es)

          phospholipid transport
          regulation of lipid metabolic process
          positive regulation of phosphatidylinositol 3-kinase activity
          inositol lipid-mediated signaling
          positive regulation of protein kinase B signaling
          positive regulation of ERK1 and ERK2 cascade
          phosphatidylinositol transporter activity
          phosphatidylinositol binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline