Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03969583_cn
          See other C1QTNF1 CNV Assays ›
          Gene Symbol
          C1QTNF1
          Assay Reference Genome
          Location

          Chr.17:79036993 on build GRCh38
          Cytoband
          17q25.3
          Assay ID Hs03969583_cn
          Size
          Availability Made To Order
          Catalog # 4400291
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Target Gene Details

          Entrez Gene ID:

          114897

          Gene Name:

          C1q and TNF related 1

          Gene Aliases:

          CTRP1, GIP, ZSIG37

          Location:

          Chr.17:79022934-79049788 on Build GRCh38

          Assay Gene Location:

          Within Intron 5
          Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
          C1QTNF1 NM_030968.5 NP_112230.1
          NM_153372.3 NP_699203.1
          NM_198593.4 NP_940995.1
          NM_198594.4 NP_940996.1
          NR_049769.3
          XM_006721664.2 XP_006721727.1
          XM_006721666.2 XP_006721729.1
          XM_024450561.2 XP_024306329.1
          XM_047435295.1 XP_047291251.1
          XM_047435296.1 XP_047291252.1
          XM_047435297.1 XP_047291253.1
          AF329840.1 AAK17964.1
          AK055541.1 BAB70947.1
          AK172767.1 BAD18750.1
          AK292034.1 BAF84723.1
          AY358424.2 AAQ88790.1
          BC021553.1 AAH21553.1
          DA875119.1
          DA877694.1
          DA910618.1
          DA938689.1
          DB126803.1
          HY047084.1

          Target Copy Number Variation Details

          DGV Version:

          Release date: 2025-12-01, GRCh GRCh38
          Target
          Variation
          Location CNV
          Subtype
          Genes
          nsv4262229 Chr.17:78653301 - 79063827 on Build GRCh38 Duplication TIMP2 CANT1 CYTH1 USP36 LGALS3BP C1QTNF1 CEP295NL C1QTNF1-AS1
          nsv521186 Chr.17:79037010 - 79049340 on Build GRCh38 Loss C1QTNF1
          nsv470610 Chr.17:78973800 - 79091723 on Build GRCh38 Loss RBFOX3 CANT1 LGALS3BP C1QTNF1 C1QTNF1-AS1 ENGASE
          nsv5533745 Chr.17:79031732 - 79038242 on Build GRCh38 Deletion C1QTNF1
          nsv6529265 Chr.17:78919705 - 79477438 on Build GRCh38 Duplication TIMP2 RBFOX3 CANT1 LGALS3BP C1QTNF1 C1QTNF1-AS1 ENGASE
          esv3641342 Chr.17:79031520 - 79038284 on Build GRCh38 Loss C1QTNF1
          nsv523650 Chr.17:78933797 - 79165563 on Build GRCh38 Loss RBFOX3 CANT1 LGALS3BP C1QTNF1 C1QTNF1-AS1 ENGASE
          nsv1057193 Chr.17:78921031 - 79481744 on Build GRCh38 Gain TIMP2 RBFOX3 CANT1 LGALS3BP C1QTNF1 C1QTNF1-AS1 ENGASE

          Back To Top

          More Information


          Additional Information:

          For this assay, SNP(s) [rs76309435] are located under a primer sequence, SNP(s) [rs115448603] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

          Set Membership:

          Intragenic Intronic Non-exonic DGV Variation Copy Number

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of cytosolic calcium ion concentration
          negative regulation of platelet activation
          positive regulation of gene expression
          negative regulation of platelet aggregation
          positive regulation of aldosterone secretion
          protein binding
          collagen binding
          identical protein binding

          Back To Top

          Related Products

          • TaqMan® Genotyping Master Mix
          • TaqPath®ProAmp® Master Mix
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® Gene Expression Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Universal PCR Master Mix, No AmpErase UNG
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-lw26s:80/100.66.78.224:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch