Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03302603_pri
          Assay Name:
          hsa-mir-92a-1
          Stem-loop ID:
          hsa-mir-19b-1 hsa-mir-20a hsa-mir-92a-1
          Mapped to miRBase Version:
          v22.1
          Chromosome Location:
          Chr.13: 91351192 - 91351278 [+] on Build GRCh38
          Stem-loop Location:
          Chr.13: 91351192 - 91351278 [+] on Build GRCh38 Chr.13: 91351065 - 91351135 [+] on Build GRCh38 Chr.13: 91351314 - 91351391 [+] on Build GRCh38
          Mature miRNA ID:
          hsa-miR-19b-3p hsa-miR-19b-1-5p hsa-miR-20a-5p hsa-miR-20a-3p hsa-miR-92a-1-5p hsa-miR-92a-3p
          Species:
          Human
          Product Type:
          TaqMan™ Pri-miRNA Assay
          Assay ID Hs03302603_pri
          Size
          Availability Made To Order
          Catalog # 4427012
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Amplicon Length:

          81

          Chromosome Location:

          Chr. 13 - 91351584 - 91351584 [-] on 91351584 Build GRCh38

          Species:

          Human

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-19b-3p MIMAT0000074
          Human hsa-miR-19b-1-5p MIMAT0004491
          Human hsa-miR-20a-5p MIMAT0000075
          Human hsa-miR-20a-3p MIMAT0004493
          Human hsa-miR-92a-1-5p MIMAT0004507
          Human hsa-miR-92a-3p MIMAT0000092

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-19b-1
          Stem-loop Accession # MI0000074
          Stem-loop Sequence
          CACUGUUCUAUGGUUAGUUUUGCAGGUUUGCAUCCAGCUGUGUGAUAUUCUGCUGUGCAAAUCCAUGCAAAACUGACUGUGGUAGUG
          Chromosome Location Chr. 13 - 91351192 - 91351278 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000074
          miRBase ID: hsa-miR-19b-3p
          Mature miRNA Sequence: UGUGCAAAUCCAUGCAAAACUGA
          Chromosome Location: Chr. 13 - 91351192 - 91351278 [+] on Build GRCh38

          miRBase Accession #: MIMAT0004491
          miRBase ID: hsa-miR-19b-1-5p
          Mature miRNA Sequence: AGUUUUGCAGGUUUGCAUCCAGC
          Chromosome Location: Chr. 13 - 91351192 - 91351278 [+] on Build GRCh38

          Gene Family ID MIPF0000011, mir-19
          Distance to Amplicon 276 bases
          Stem-loop ID hsa-mir-20a
          Stem-loop Accession # MI0000076
          Stem-loop Sequence
          GUAGCACUAAAGUGCUUAUAGUGCAGGUAGUGUUUAGUUAUCUACUGCAUUAUGAGCACUUAAAGUACUGC
          Chromosome Location Chr. 13 - 91351065 - 91351135 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000075
          miRBase ID: hsa-miR-20a-5p
          Mature miRNA Sequence: UAAAGUGCUUAUAGUGCAGGUAG
          Chromosome Location: Chr. 13 - 91351065 - 91351135 [+] on Build GRCh38

          miRBase Accession #: MIMAT0004493
          miRBase ID: hsa-miR-20a-3p
          Mature miRNA Sequence: ACUGCAUUAUGAGCACUUAAAG
          Chromosome Location: Chr. 13 - 91351065 - 91351135 [+] on Build GRCh38

          Gene Family ID MIPF0000001, mir-17
          Distance to Amplicon 419 bases
          Stem-loop ID hsa-mir-92a-1
          Stem-loop Accession # MI0000093
          Stem-loop Sequence
          CUUUCUACACAGGUUGGGAUCGGUUGCAAUGCUGUGUUUCUGUAUGGUAUUGCACUUGUCCCGGCCUGUUGAGUUUGG
          Chromosome Location Chr. 13 - 91351314 - 91351391 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0004507
          miRBase ID: hsa-miR-92a-1-5p
          Mature miRNA Sequence: AGGUUGGGAUCGGUUGCAAUGCU
          Chromosome Location: Chr. 13 - 91351314 - 91351391 [+] on Build GRCh38

          miRBase Accession #: MIMAT0000092
          miRBase ID: hsa-miR-92a-3p
          Mature miRNA Sequence: UAUUGCACUUGUCCCGGCCUGU
          Chromosome Location: Chr. 13 - 91351314 - 91351391 [+] on Build GRCh38

          Gene Family ID MIPF0000013, mir-25
          Distance to Amplicon 163 bases

          Back To Top

          More Information




          Related Products

          Ambion® Anti-miR™ miRNA Inhibitor : AM10057
          Ambion® Pre-miR™ miRNA Precursor : PM10057
          TaqMan™ MicroRNA Assay : 000580
          mirVana® miRNA inhibitor : MH10057
          mirVana® miRNA mimic : MC10057
          TaqMan™ Advanced miRNA Assay : 478586_mir
          TaqMan™ Advanced miRNA Assay : rno478586_mir
          Ambion® Anti-miR™ miRNA Inhibitor : AM12696
          Ambion® Pre-miR™ miRNA Precursor : PM12696
          TaqMan™ MicroRNA Assay : 002137
          mirVana® miRNA inhibitor : MH12696
          mirVana® miRNA mimic : MC12696
          TaqMan™ Advanced miRNA Assay : 479205_mir
          Ambion® Anti-miR™ miRNA Inhibitor : AM12903
          Ambion® Pre-miR™ miRNA Precursor : PM12903
          TaqMan™ MicroRNA Assay : 002437
          mirVana® miRNA inhibitor : MH12903
          mirVana® miRNA mimic : MC12903
          TaqMan™ Advanced miRNA Assay : 478317_mir
          Ambion® Anti-miR™ miRNA Inhibitor : AM10916
          Ambion® Pre-miR™ miRNA Precursor : PM10916
          TaqMan™ MicroRNA Assay : 000431
          mirVana® miRNA inhibitor : MH10916
          mirVana® miRNA mimic : MC10916
          TaqMan™ Advanced miRNA Assay : 477827_mir
          Ambion® Anti-miR™ miRNA Inhibitor : AM10629
          Ambion® Pre-miR™ miRNA Precursor : PM10629
          TaqMan™ MicroRNA Assay : 000396
          mirVana® miRNA inhibitor : MH10629
          mirVana® miRNA mimic : MC10629
          TaqMan™ Advanced miRNA Assay : 478264_mir
          TaqMan™ Advanced miRNA Assay : mmu478264_mir
          TaqMan™ Advanced miRNA Assay : rno478264_mir
          Ambion® Anti-miR™ miRNA Inhibitor : AM13042
          Ambion® Pre-miR™ miRNA Precursor : PM13042
          TaqMan™ MicroRNA Assay : 002425
          mirVana® miRNA inhibitor : MH13042
          mirVana® miRNA mimic : MC13042
          TaqMan™ Advanced miRNA Assay : mmu481634_mir
          TaqMan™ Advanced miRNA Assay : 477962_mir


          miRBase Alias:

          Human: hsa-mir-19b-1 (v21)
          Human: hsa-mir-20a (v21)

          Back To Top

          Related Products

          • High Capacity RNA-to-cDNA Kit
          • SuperScript® IV VILO™ Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-7kv6q:80/100.66.72.150:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch