Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MH13042
          Assay Name: hsa-miR-19b-1-5p
          miRBase Accession Number: MI0000074
          miRBase Version: v22.1
          Chromosome Location: Chr.13: 91351192 - 91351278 [+] on Build GRCh38
          Mature miRNA Sequence: AGUUUUGCAGGUUUGCAUCCAGC
          Species: Human, Mouse, Alligator mississippiensis, Chicken, Columba livia, Cricetulus griseus, Dasypus novemcinctus, Goat, Guinea pig, Monodelphis domestica, Ophiophagus hannah, Ornithorhynchus anatinus, Pteropus alecto, Rabbit, Taeniopygia guttata
          Product Type: mirVana® miRNA inhibitor
          Assay ID MH13042

          Purification
          Size
          Availability Made To Order
          Catalog # 4464084
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000011, mir-19

          Mature miRNA Sequence:

          AGUUUUGCAGGUUUGCAUCCAGC

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-19b-1-5p MIMAT0004491
          Mouse mmu-miR-19b-1-5p MIMAT0017065
          Alligator mississippiensis ami-miR-19b-5p MIMAT0038052
          Chicken gga-miR-19b-5p MIMAT0026496
          Columba livia cli-miR-19b-5p MIMAT0038429
          Cricetulus griseus cgr-miR-19b-5p MIMAT0023840
          View More Rows
          Dasypus novemcinctus dno-miR-19b-5p MIMAT0047517
          Goat chi-miR-19b-5p MIMAT0036043
          Guinea pig cpo-miR-19b-5p MIMAT0046865
          Monodelphis domestica mdo-miR-19b-1-5p MIMAT0026708
          Ophiophagus hannah oha-miR-19b-5p MIMAT0036797
          Ornithorhynchus anatinus oan-miR-19b-2-5p MIMAT0031104
          Pteropus alecto pal-miR-19-2-5p MIMAT0040168
          Rabbit ocu-miR-19b-5p MIMAT0048117
          Taeniopygia guttata tgu-miR-19b-1-5p MIMAT0026981

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-19b-1
          Stem-loop Accession # MI0000074
          Stem-loop Sequence
          CACUGUUCUAUGGUUAGUUUUGCAGGUUUGCAUCCAGCUGUGUGAUAUUCUGCUGUGCAAAUCCAUGCAAAACUGACUGUGGUAGUG
          Chromosome Location Chr. 13 - 91351192 - 91351278 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0004491
          miRBase ID: hsa-miR-19b-1-5p
          Mature miRNA Sequence: AGUUUUGCAGGUUUGCAUCCAGC
          Chromosome Location: Chr. 13 - 91351192 - 91351278 [+] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-19b-1
          Stem-loop Accession # MI0000718
          Stem-loop Sequence
          CACUGGUCUAUGGUUAGUUUUGCAGGUUUGCAUCCAGCUGUAUAAUAUUCUGCUGUGCAAAUCCAUGCAAAACUGACUGUGGUGGUG
          Chromosome Location Chr. 14 - 115044305 - 115044391 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0017065
          miRBase ID: mmu-miR-19b-1-5p
          Mature miRNA Sequence: AGUUUUGCAGGUUUGCAUCCAGC
          Chromosome Location: Chr. 14 - 115044305 - 115044391 [+] on Build GRCm38

          Alligator mississippiensis

          Stem-loop ID ami-mir-19b
          Stem-loop Accession # MI0029675
          Stem-loop Sequence
          CACUGUUCUCUGGUUAGUUUUGCAGGUUUGCAUCCAGCUGUAUGAUACUCUGCUGUGCAAAUCCAUGCAAAACUGACUGUGGCAGUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038052
          miRBase ID: ami-miR-19b-5p
          Mature miRNA Sequence: AGUUUUGCAGGUUUGCAUCCAGC
          Chromosome Location: NA

          Chicken

          Stem-loop ID gga-mir-19b
          Stem-loop Accession # MI0001180
          Stem-loop Sequence
          CACUGUUCUCUGGUUAGUUUUGCAGGUUUGCAUCCAGCUGUAUGAUACUCUGCUGUGCAAAUCCAUGCAAAACUGACUGUGGCAGUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026496
          miRBase ID: gga-miR-19b-5p
          Mature miRNA Sequence: AGUUUUGCAGGUUUGCAUCCAGC
          Chromosome Location: NA

          Columba livia

          Stem-loop ID cli-mir-19b-1
          Stem-loop Accession # MI0029916
          Stem-loop Sequence
          CUCUGGUUAGUUUUGCAGGUUUGCAUCCAGCUGUAUGAUACUCUGCUGUGCAAAUCCAUGCAAAACUGACUGUGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038429
          miRBase ID: cli-miR-19b-5p
          Mature miRNA Sequence: AGUUUUGCAGGUUUGCAUCCAGC
          Chromosome Location: NA

          Cricetulus griseus

          Stem-loop ID cgr-mir-19b
          Stem-loop Accession # MI0020451
          Stem-loop Sequence
          GUCUAUGGUUAGUUUUGCAGGUUUGCAUCCAGCUGUAUAAUACUCUGCUGUGCAAAUCCAUGCAAAACUGACUGUGGUGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0023840
          miRBase ID: cgr-miR-19b-5p
          Mature miRNA Sequence: AGUUUUGCAGGUUUGCAUCCAGC
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-19b-1
          Stem-loop Accession # MI0038913
          Stem-loop Sequence
          AGUUUUGCAGGUUUGCAUCCAGCUGUGUGAUAUUCUGCUGUGCAAAUCCAUGCAAAACUGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047517
          miRBase ID: dno-miR-19b-5p
          Mature miRNA Sequence: AGUUUUGCAGGUUUGCAUCCAGC
          Chromosome Location: NA

          Goat

          Stem-loop ID chi-mir-19b
          Stem-loop Accession # MI0030678
          Stem-loop Sequence
          CACUGUUCUAUGGUUAGUUUUGCAGGUUUGCAUCCAGCUGUGUGAUAUUCUGCUGUGCAAAUCCAUGCAAAACUGACUGUGGUAGUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036043
          miRBase ID: chi-miR-19b-5p
          Mature miRNA Sequence: AGUUUUGCAGGUUUGCAUCCAGC
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-19b-1
          Stem-loop Accession # MI0038569
          Stem-loop Sequence
          AGUUUUGCAGGUUUGCAUCCAGCUGUGUGAUAUUCUGCUGUGCAAAUCCAUGCAAAACUGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046865
          miRBase ID: cpo-miR-19b-5p
          Mature miRNA Sequence: AGUUUUGCAGGUUUGCAUCCAGC
          Chromosome Location: NA

          Monodelphis domestica

          Stem-loop ID mdo-mir-19b-1
          Stem-loop Accession # MI0005358
          Stem-loop Sequence
          GCACUGUUCUAUGGUUAGUUUUGCAGGUUUGCAUCCAGCUGUAUGAUAUUCUGCUGUGCAAAUCCAUGCAAAACUGACUGUGGUGGUGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026708
          miRBase ID: mdo-miR-19b-1-5p
          Mature miRNA Sequence: AGUUUUGCAGGUUUGCAUCCAGC
          Chromosome Location: NA

          Ophiophagus hannah

          Stem-loop ID oha-mir-19b
          Stem-loop Accession # MI0031396
          Stem-loop Sequence
          UAAACACUGUUCUCUGGUUAGUUUUGCAGGUUUGCAUCCAGCUGUAUGAUACUCUGCUGUGCAAAUCCAUGCAAAACUGACUGUGGCAGUGAAAAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036797
          miRBase ID: oha-miR-19b-5p
          Mature miRNA Sequence: AGUUUUGCAGGUUUGCAUCCAGC
          Chromosome Location: NA

          Ornithorhynchus anatinus

          Stem-loop ID oan-mir-19b-2
          Stem-loop Accession # MI0024088
          Stem-loop Sequence
          AGUUUUGCAGGUUUGCAUCCAGCUGUAUGGUACUCUGCUGUGCAAAUCCAUGCAAAACUGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0031104
          miRBase ID: oan-miR-19b-2-5p
          Mature miRNA Sequence: AGUUUUGCAGGUUUGCAUCCAGC
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-19-2
          Stem-loop Accession # MI0032530
          Stem-loop Sequence
          AGUUUUGCAGGUUUGCAUCCAGCUGUGUGAUAUUCUGCUGUGCAAAUCCAUGCAAAACUGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0040168
          miRBase ID: pal-miR-19-2-5p
          Mature miRNA Sequence: AGUUUUGCAGGUUUGCAUCCAGC
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-19b-1
          Stem-loop Accession # MI0039229
          Stem-loop Sequence
          AGUUUUGCAGGUUUGCAUCCAGCUGUGUGAUAUUCUGCUGUGCAAAUCCAUGCAAAACUGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048117
          miRBase ID: ocu-miR-19b-5p
          Mature miRNA Sequence: AGUUUUGCAGGUUUGCAUCCAGC
          Chromosome Location: NA

          Taeniopygia guttata

          Stem-loop ID tgu-mir-19b-1
          Stem-loop Accession # MI0013719
          Stem-loop Sequence
          UUAGUUUUGCAGGUUUGCAUCCAGCUGUAUGAUACUCUGCUGUGCAAAUCCAUGCAAAACUGACUGUGGCAGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026981
          miRBase ID: tgu-miR-19b-1-5p
          Mature miRNA Sequence: AGUUUUGCAGGUUUGCAUCCAGC
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03302603_pri
          TaqMan™ Pri-miRNA Assay : Mm03306814_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM13042
          Ambion® Pre-miR™ miRNA Precursor : PM13042
          TaqMan™ MicroRNA Assay : 002425
          mirVana® miRNA mimic : MC13042
          TaqMan™ Advanced miRNA Assay : mmu481634_mir
          TaqMan™ Advanced miRNA Assay : 477962_mir


          miRBase Alias:

          Human: hsa-miR-19b-1* (v17)
          Mouse: mmu-miR-19b-1* (v17)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-lw26s:80/100.66.78.224:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch