Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
CD4
Transcript:
NM_001195015.2
Assay Name:
Mutation genome location:
chr.12 6799989-6800488 on build GRCh38
Assay ID
Hs00115688_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Between 400-700 bp
Gene Ontology Categories:
Function(s)
Process(es)
cytokine production
adaptive immune response
induction by virus of host cell-cell fusion
immune response
signal transduction
cell surface receptor signaling pathway
enzyme linked receptor protein signaling pathway
transmembrane receptor protein tyrosine kinase signaling pathway
positive regulation of calcium ion transport into cytosol
fusion of virus membrane with host plasma membrane
T cell differentiation
entry into host cell
T cell costimulation
response to estradiol
maintenance of protein location in cell
response to vitamin D
helper T cell enhancement of adaptive immune response
positive regulation of T cell proliferation
T cell selection
positive regulation of interleukin-2 biosynthetic process
positive regulation of protein kinase activity
regulation of defense response to virus by virus
positive regulation of peptidyl-tyrosine phosphorylation
defense response to Gram-negative bacterium
positive regulation of calcium-mediated signaling
T cell receptor signaling pathway
regulation of T cell activation
virus receptor activity
glycoprotein binding
receptor activity
transmembrane signaling receptor activity
extracellular matrix structural constituent
protein binding
zinc ion binding
coreceptor activity
immunoglobulin binding
enzyme binding
protein kinase binding
MHC class II protein binding
protein homodimerization activity
protein tyrosine kinase binding
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-5cvsx:80/100.66.79.163:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline