Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
MAPK3
Transcript:
NM_002746.2
Assay Name:
Mutation genome location:
chr.16 30117969-30118470 on build GRCh38
Assay ID
Hs00154479_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Between 400-700 bp
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
MAPK cascade
activation of MAPK activity
MAPK import into nucleus
positive regulation of protein phosphorylation
transcription initiation from RNA polymerase I promoter
protein complex assembly
protein phosphorylation
apoptotic process
DNA damage induced protein phosphorylation
cell cycle
axon guidance
fibroblast growth factor receptor signaling pathway
response to toxic substance
regulation of phosphatidylinositol 3-kinase signaling
viral process
phosphorylation
peptidyl-serine phosphorylation
sensory perception of pain
arachidonic acid metabolic process
platelet activation
BMP signaling pathway
thyroid gland development
positive regulation of cyclase activity
lipopolysaccharide-mediated signaling pathway
positive regulation of telomere maintenance via telomerase
regulation of stress-activated MAPK cascade
positive regulation of histone phosphorylation
positive regulation of histone acetylation
peptidyl-tyrosine autophosphorylation
Fc-epsilon receptor signaling pathway
Fc-gamma receptor signaling pathway involved in phagocytosis
outer ear morphogenesis
response to exogenous dsRNA
positive regulation of translation
positive regulation of transcription from RNA polymerase II promoter
thymus development
regulation of sequence-specific DNA binding transcription factor activity
cartilage development
regulation of cytoskeleton organization
positive regulation of telomerase activity
Bergmann glial cell differentiation
face development
JAK-STAT cascade involved in growth hormone signaling pathway
lung morphogenesis
trachea formation
cardiac neural crest cell development involved in heart development
ERK1 and ERK2 cascade
positive regulation of ERK1 and ERK2 cascade
interleukin-1-mediated signaling pathway
response to epidermal growth factor
cellular response to mechanical stimulus
caveolin-mediated endocytosis
regulation of Golgi inheritance
regulation of cellular response to heat
positive regulation of telomere capping
positive regulation of xenophagy
regulation of early endosome to late endosome transport
negative regulation of apolipoprotein binding
phosphotyrosine binding
protein serine/threonine kinase activity
MAP kinase activity
protein binding
ATP binding
kinase activity
phosphatase binding
scaffold protein binding
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-5cvsx:80/100.66.79.163:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline