Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
SNCA
Transcript:
NM_001146054.1
Assay Name:
Mutation genome location:
chr.4 89822105-89822644 on build GRCh38
Assay ID
Hs00251569_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Between 400-700 bp
Panther Classification:
Molecular Function -
membrane trafficking regulatory protein
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
microglial cell activation
positive regulation of receptor recycling
negative regulation of protein phosphorylation
positive regulation of neurotransmitter secretion
fatty acid metabolic process
neutral lipid metabolic process
phospholipid metabolic process
apoptotic process
activation of cysteine-type endopeptidase activity involved in apoptotic process
mitochondrial membrane organization
aging
adult locomotory behavior
response to iron(II) ion
regulation of phospholipase activity
negative regulation of platelet-derived growth factor receptor signaling pathway
regulation of glutamate secretion
regulation of dopamine secretion
negative regulation of microtubule polymerization
receptor internalization
protein destabilization
response to magnesium ion
negative regulation of transporter activity
response to lipopolysaccharide
negative regulation of monooxygenase activity
positive regulation of peptidyl-serine phosphorylation
response to interferon-gamma
cellular response to oxidative stress
negative regulation of histone acetylation
regulation of locomotion
dopamine biosynthetic process
response to drug
mitochondrial ATP synthesis coupled electron transport
regulation of macrophage activation
positive regulation of apoptotic process
negative regulation of apoptotic process
negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
extracellular fibril organization
negative regulation of neuron apoptotic process
cellular protein metabolic process
cellular response to fibroblast growth factor stimulus
positive regulation of endocytosis
negative regulation of exocytosis
negative regulation of dopamine metabolic process
behavioral response to cocaine
regulation of long-term neuronal synaptic plasticity
synaptic vesicle endocytosis
synapse organization
regulation of acyl-CoA biosynthetic process
positive regulation of release of sequestered calcium ion into cytosol
dopamine uptake involved in synaptic transmission
negative regulation of dopamine uptake involved in synaptic transmission
negative regulation of serotonin uptake
negative regulation of norepinephrine uptake
calcium ion homeostasis
oxidation-reduction process
excitatory postsynaptic potential
long-term synaptic potentiation
positive regulation of inositol phosphate biosynthetic process
negative regulation of thrombin receptor signaling pathway
response to interleukin-1
cellular response to copper ion
cellular response to epinephrine stimulus
positive regulation of protein serine/threonine kinase activity
negative regulation of neuron death
negative regulation of mitochondrial electron transport, NADH to ubiquinone
positive regulation of glutathione peroxidase activity
positive regulation of hydrogen peroxide catabolic process
regulation of synaptic vesicle recycling
regulation of reactive oxygen species biosynthetic process
negative regulation of chaperone-mediated autophagy
magnesium ion binding
fatty acid binding
copper ion binding
calcium ion binding
protein binding
phospholipid binding
microtubule binding
ferrous iron binding
zinc ion binding
oxidoreductase activity
kinesin binding
protein domain specific binding
histone binding
identical protein binding
alpha-tubulin binding
cysteine-type endopeptidase inhibitor activity involved in apoptotic process
phospholipase binding
transcription regulatory region DNA binding
dynein binding
protein N-terminus binding
tau protein binding
beta-tubulin binding
phosphoprotein binding
phospholipase D inhibitor activity
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-pgj68:80/100.66.79.163:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline