Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
LRRK2
Transcript:
NM_198578.3
Assay Name:
Mutation genome location:
chr.12 40263549-40264055 on build GRCh38
Assay ID
Hs00335653_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Between 400-700 bp
Panther Classification:
Molecular Function -
scaffold/adaptor protein
Gene Ontology Categories:
Function(s)
Process(es)
MAPK cascade
activation of MAPKK activity
activation of MAPK activity
negative regulation of protein phosphorylation
positive regulation of protein phosphorylation
protein phosphorylation
endocytosis
autophagy
response to oxidative stress
mitochondrion organization
Golgi organization
lysosome organization
small GTPase mediated signal transduction
neuromuscular junction development
determination of adult lifespan
cellular response to starvation
regulation of autophagy
positive regulation of autophagy
regulation of protein kinase A signaling
negative regulation of protein processing
regulation of neuron maturation
negative regulation of macroautophagy
phosphorylation
peptidyl-serine phosphorylation
peptidyl-threonine phosphorylation
calcium-mediated signaling
olfactory bulb development
tangential migration from the subventricular zone to the olfactory bulb
positive regulation of protein ubiquitination
negative regulation of protein binding
positive regulation of protein binding
positive regulation of proteasomal ubiquitin-dependent protein catabolic process
negative regulation of GTPase activity
cellular response to oxidative stress
cellular protein localization
intracellular signal transduction
regulation of kidney size
exploration behavior
locomotory exploration behavior
regulation of lysosomal lumen pH
regulation of locomotion
regulation of membrane potential
positive regulation of programmed cell death
positive regulation of MAP kinase activity
positive regulation of GTPase activity
GTP metabolic process
protein autophosphorylation
intracellular distribution of mitochondria
neuron projection morphogenesis
regulation of calcium-mediated signaling
mitochondrion localization
regulation of mitochondrial depolarization
regulation of synaptic transmission, glutamatergic
canonical Wnt signaling pathway
excitatory postsynaptic potential
regulation of dopamine receptor signaling pathway
positive regulation of dopamine receptor signaling pathway
regulation of canonical Wnt signaling pathway
regulation of dendritic spine morphogenesis
protein localization to mitochondrion
neuron death
cellular response to manganese ion
reactive oxygen species metabolic process
regulation of mitochondrial fission
positive regulation of canonical Wnt signaling pathway
negative regulation of excitatory postsynaptic potential
regulation of neuron death
negative regulation of neuron death
negative regulation of endoplasmic reticulum stress-induced intrinsic apoptotic signaling pathway
positive regulation of protein autoubiquitination
regulation of neuroblast proliferation
regulation of synaptic vesicle transport
negative regulation of late endosome to lysosome transport
negative regulation of autophagosome assembly
negative regulation of thioredoxin peroxidase activity by peptidyl-threonine phosphorylation
negative regulation of hydrogen peroxide-induced cell death
negative regulation of protein targeting to mitochondrion
negative regulation of protein processing involved in protein targeting to mitochondrion
cellular response to dopamine
Wnt signalosome assembly
regulation of retrograde transport, endosome to Golgi
regulation of branching morphogenesis of a nerve
regulation of synaptic vesicle exocytosis
negative regulation of peroxidase activity
SNARE binding
glycoprotein binding
actin binding
GTPase activity
protein kinase activity
protein serine/threonine kinase activity
MAP kinase kinase activity
GTPase activator activity
protein binding
ATP binding
GTP binding
microtubule binding
tubulin binding
kinase activity
Rho GTPase binding
syntaxin-1 binding
receptor signaling complex scaffold activity
clathrin binding
GTP-dependent protein kinase activity
peroxidase inhibitor activity
co-receptor binding
identical protein binding
protein homodimerization activity
ion channel binding
protein kinase A binding
beta-catenin destruction complex binding
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline