Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
MRE11A
Transcript:
Assay Name:
Mutation genome location:
chr.11 94418616-94419204 on build GRCh38
Assay ID
Hs00429126_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Between 400-700 bp
Gene Ontology Categories:
Function(s)
Process(es)
regulation of mitotic recombination
double-strand break repair via homologous recombination
DNA double-strand break processing
DNA synthesis involved in DNA repair
strand displacement
DNA replication
DNA repair
double-strand break repair
double-strand break repair via nonhomologous end joining
DNA recombination
cellular response to DNA damage stimulus
telomere maintenance via telomerase
sister chromatid cohesion
mitotic G2 DNA damage checkpoint
synapsis
reciprocal meiotic recombination
heart development
cell proliferation
viral process
intra-S DNA damage checkpoint
telomeric 3' overhang formation
positive regulation of protein autophosphorylation
positive regulation of telomere maintenance
positive regulation of type I interferon production
DNA duplex unwinding
negative regulation of DNA endoreduplication
positive regulation of kinase activity
negative regulation of apoptotic process
negative regulation of viral entry into host cell
nucleic acid phosphodiester bond hydrolysis
cell-cell adhesion
regulation of signal transduction by p53 class mediator
single-stranded DNA endodeoxyribonuclease activity
DNA binding
double-stranded DNA binding
ATP-dependent DNA helicase activity
nuclease activity
endodeoxyribonuclease activity
protein binding
protein C-terminus binding
3'-5' exonuclease activity
manganese ion binding
cadherin binding involved in cell-cell adhesion
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-5cvsx:80/100.66.79.163:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline