Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
PML
Transcript:
NM_033240.2
Assay Name:
Mutation genome location:
chr.15 73998179-73998339 on build GRCh38
Assay transcription exon location:
Within Exon 2
Assay ID
Hs00522099_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Less than 200 bp
Gene Ontology Categories:
Function(s)
Process(es)
response to hypoxia
regulation of protein phosphorylation
transcription, DNA-templated
regulation of transcription, DNA-templated
protein complex assembly
activation of cysteine-type endopeptidase activity involved in apoptotic process
DNA damage response, signal transduction by p53 class mediator resulting in cell cycle arrest
cell cycle arrest
transforming growth factor beta receptor signaling pathway
common-partner SMAD protein phosphorylation
SMAD protein import into nucleus
negative regulation of cell proliferation
intrinsic apoptotic signaling pathway in response to oxidative stress
response to UV
response to gamma radiation
regulation of calcium ion transport into cytosol
fibroblast migration
negative regulation of angiogenesis
protein sumoylation
myeloid cell differentiation
regulation of cell adhesion
negative regulation of cell growth
PML body organization
positive regulation of telomere maintenance
endoplasmic reticulum calcium ion homeostasis
circadian regulation of gene expression
response to cytokine
regulation of circadian rhythm
intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
entrainment of circadian clock by photoperiod
proteasome-mediated ubiquitin-dependent protein catabolic process
innate immune response
cell fate commitment
regulation of MHC class I biosynthetic process
negative regulation of transcription, DNA-templated
positive regulation of fibroblast proliferation
retinoic acid receptor signaling pathway
negative regulation of interleukin-1 beta secretion
maintenance of protein location in nucleus
defense response to virus
interferon-gamma-mediated signaling pathway
branching involved in mammary gland duct morphogenesis
intrinsic apoptotic signaling pathway in response to endoplasmic reticulum stress
cellular response to interleukin-4
cellular senescence
extrinsic apoptotic signaling pathway
regulation of signal transduction by p53 class mediator
negative regulation of viral release from host cell
negative regulation of protein ubiquitination involved in ubiquitin-dependent protein catabolic process
regulation of double-strand break repair
positive regulation of extrinsic apoptotic signaling pathway
DNA binding
protein binding
zinc ion binding
ubiquitin protein ligase binding
SUMO binding
protein homodimerization activity
SMAD binding
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline