Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
CIB1
Transcript:
NM_006384.3
Assay Name:
Mutation genome location:
chr.15 90232132-90232401 on build GRCh38
Assay ID
Hs00616648_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Between 200-400 bp
Gene Ontology Categories:
Function(s)
Process(es)
angiogenesis
negative regulation of protein phosphorylation
positive regulation of protein phosphorylation
positive regulation of cell-matrix adhesion
response to ischemia
double-strand break repair
apoptotic process
cellular response to DNA damage stimulus
negative regulation of microtubule depolymerization
endomitotic cell cycle
cell adhesion
spermatid development
positive regulation of cell proliferation
negative regulation of cell proliferation
negative regulation of neuron projection development
platelet formation
positive regulation of cell growth
positive regulation of cell migration
cytoplasmic microtubule organization
positive regulation of cell adhesion mediated by integrin
thrombopoietin-mediated signaling pathway
regulation of cell proliferation
negative regulation of apoptotic process
positive regulation of catalytic activity
negative regulation of megakaryocyte differentiation
positive regulation of NF-kappaB transcription factor activity
cell division
regulation of cell division
negative regulation of protein kinase B signaling
positive regulation of ERK1 and ERK2 cascade
positive regulation of calcineurin-NFAT signaling cascade
cellular response to tumor necrosis factor
cellular response to growth factor stimulus
negative regulation of protein serine/threonine kinase activity
positive regulation of protein serine/threonine kinase activity
positive regulation of establishment of protein localization to plasma membrane
positive regulation of cell migration involved in sprouting angiogenesis
positive regulation of protein targeting to membrane
extrinsic apoptotic signaling pathway
positive regulation of substrate adhesion-dependent cell spreading
cellular response to nerve growth factor stimulus
positive regulation of male germ cell proliferation
calcium ion binding
protein binding
protein C-terminus binding
calcium-dependent protein kinase inhibitor activity
Ras GTPase binding
protein kinase binding
protein serine/threonine kinase inhibitor activity
protein anchor
ion channel binding
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline