Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
C2orf88
Transcript:
NM_005259.2
Assay Name:
Mutation genome location:
chr.2 190062089-190062326 on build GRCh38
Gene:
MSTN
Transcript:
NM_005259.2
Assay Name:
Mutation genome location:
chr.2 190062089-190062326 on build GRCh38
Assay ID
Hs00795204_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Between 200-400 bp
Panther Classification:
Molecular Function -
scaffold/adaptor protein
growth factor
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of defense response to virus by host
transforming growth factor beta receptor signaling pathway
muscle organ development
response to heat
response to gravity
positive regulation of pathway-restricted SMAD protein phosphorylation
skeletal muscle atrophy
negative regulation of muscle hypertrophy
response to muscle activity
ovulation cycle process
response to testosterone
negative regulation of kinase activity
regulation of apoptotic process
skeletal muscle tissue regeneration
regulation of MAPK cascade
response to estrogen
response to ethanol
negative regulation of myoblast differentiation
positive regulation of transcription, DNA-templated
negative regulation of insulin receptor signaling pathway
muscle cell cellular homeostasis
cell development
negative regulation of skeletal muscle tissue growth
response to electrical stimulus
negative regulation of protein kinase B signaling
SMAD protein signal transduction
cellular response to dexamethasone stimulus
mitophagy in response to mitochondrial depolarization
xenophagy
negative regulation of skeletal muscle satellite cell proliferation
negative regulation of satellite cell differentiation
negative regulation of myoblast proliferation
protein binding
protein kinase A regulatory subunit binding
receptor binding
cytokine activity
transforming growth factor beta receptor binding
growth factor activity
heparin binding
identical protein binding
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-5cvsx:80/100.66.79.163:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline