Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
NF1
Transcript:
NM_001042492.2
Assay Name:
Mutation genome location:
chr.17 31223393-31223622 on build GRCh38
Assay ID
Hs00811656_CE
Purification
DS
HPLC
Size
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
25 nmol
25 nmol
50 nmol
50 nmol
Availability
Made To Order
Catalog #
A15629
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Set Membership:
Between 200-400 bp
Panther Classification:
Molecular Function -
GTPase-activating protein
Gene Ontology Categories:
Function(s)
Process(es)
MAPK cascade
osteoblast differentiation
metanephros development
response to hypoxia
liver development
negative regulation of endothelial cell proliferation
positive regulation of endothelial cell proliferation
regulation of cell-matrix adhesion
negative regulation of cell-matrix adhesion
negative regulation of protein kinase activity
cell communication
Ras protein signal transduction
negative regulation of neuroblast proliferation
brain development
peripheral nervous system development
heart development
skeletal muscle tissue development
visual learning
extrinsic apoptotic signaling pathway via death domain receptors
regulation of gene expression
Schwann cell development
phosphatidylinositol 3-kinase signaling
negative regulation of angiogenesis
spinal cord development
amygdala development
forebrain astrocyte development
neural tube development
cerebral cortex development
myelination in peripheral nervous system
actin cytoskeleton organization
extracellular matrix organization
collagen fibril organization
adrenal gland development
negative regulation of cell migration
regulation of synaptic transmission, GABAergic
cellular response to heat
negative regulation of Rac protein signal transduction
wound healing
negative regulation of transcription factor import into nucleus
positive regulation of apoptotic process
regulation of GTPase activity
negative regulation of MAP kinase activity
negative regulation of MAPK cascade
pigmentation
positive regulation of neuron apoptotic process
regulation of blood vessel endothelial cell migration
positive regulation of GTPase activity
regulation of bone resorption
negative regulation of osteoclast differentiation
regulation of glial cell differentiation
positive regulation of adenylate cyclase activity
regulation of angiogenesis
negative regulation of Ras protein signal transduction
negative regulation of neurotransmitter secretion
negative regulation of fibroblast proliferation
regulation of long-term neuronal synaptic plasticity
sympathetic nervous system development
camera-type eye morphogenesis
negative regulation of astrocyte differentiation
negative regulation of oligodendrocyte differentiation
smooth muscle tissue development
artery morphogenesis
forebrain morphogenesis
cognition
gamma-aminobutyric acid secretion, neurotransmission
glutamate secretion, neurotransmission
observational learning
regulation of long-term synaptic potentiation
positive regulation of extrinsic apoptotic signaling pathway via death domain receptors
positive regulation of extrinsic apoptotic signaling pathway in absence of ligand
GTPase activator activity
protein binding
phosphatidylethanolamine binding
phosphatidylcholine binding
Back To Top
Related Products
POP-6
™
Polymer
POP-7
™
Polymer
BigDye
®
Terminator v1.1 Cycle Sequencing Kit
BigDye
®
Terminator v3.1 Cycle Sequencing Kit
BigDye
®
Direct Cycle Sequencing Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.46.0-Offline