Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prr18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178774.4 | NP_848889.3 | 2 | 904 | ||
GenBank mRNA | AK035939.1 | - | 1 | 727 | ||
AK039752.1 | - | 2 | 729 | |||
AK045379.1 | - | 2 | 1036 | |||
BC060190.1 | - | 2 | 1060 | 6833275 |