Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc9b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178877.6 | NP_849208.4 | 12 | 1684 | ||
GenBank mRNA | AK029417.1 | - | 9 | 1235 | ||
AK039655.1 | - | 3 | 683 | |||
BC090977.1 | - | 12 | 1637 | 30503721 |