Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sbf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177324.2 | NP_796298.2 | 6 | 702 | ||
GenBank mRNA | AK046622.1 | - | 6 | 690 | ||
AK048952.1 | - | 6 | 679 | |||
AK080952.1 | - | 6 | 703 | |||
AK080963.1 | - | 6 | 697 | |||
AK087396.1 | - | 6 | 697 | |||
AK173257.1 | - | 6 | 661 | |||
BC048706.1 | - | 6 | 687 | |||
BC067204.1 | - | 7 | 740 |