Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcorl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178782.4 | NP_848897.3 | 5 | 1993 | ||
GenBank mRNA | AK081004.1 | - | 1 | 561 |