Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp956 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178898.4 | NP_849229.2 | 5 | 1104 | ||
GenBank mRNA | AK040541.1 | - | 5 | 1170 | ||
AK045736.1 | - | 5 | 1093 | |||
AK133549.1 | - | 5 | 1105 | |||
AK162106.1 | - | 5 | 1031 | |||
BC090657.1 | - | 5 | 1046 | |||
BC096536.1 | - | 4 | 814 | |||
BC119280.1 | - | 5 | 982 | 8734029 | ||
BC120581.1 | - | 5 | 983 | 40129504 | ||
BC144752.1 | - | 5 | 983 | 9053300 |