Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr107 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178760.4 | NP_848875.2 | 18 | 2156 | ||
GenBank mRNA | AK030433.1 | - | 18 | 2152 | ||
AK034815.1 | - | 18 | 2139 | |||
AK082237.1 | - | 18 | 2154 | |||
AK129407.1 | - | 18 | 2051 | |||
AK154530.1 | - | 18 | 2152 | |||
BC052649.1 | - | 1 | 109 | |||
BC055799.1 | - | 1 | 269 | |||
BC117900.1 | - | 18 | 2134 | 40097045 | ||
BC117901.1 | - | 18 | 2134 | 40097052 |
Set Membership: |
G-protein coupled receptor |