Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pigb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK153819.1 | - | 13 | 2735 |
Gene Symbol: LOC100135765 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK034716.1 | - | 1 | 1542 | ||
AK035452.1 | - | 1 | 680 |